Skip to Content
Merck
All Photos(2)

Key Documents

674974

Sigma-Aldrich

6-Mercaptohexanoic acid

90%

Synonym(s):

MHA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12O2S
CAS Number:
Molecular Weight:
148.22
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Assay

90%

refractive index

n20/D 1.4846

density

1.0715 g/mL at 25 °C

storage temp.

−20°C

SMILES string

OC(=O)CCCCCS

InChI

1S/C6H12O2S/c7-6(8)4-2-1-3-5-9/h9H,1-5H2,(H,7,8)

InChI key

CMNQZZPAVNBESS-UHFFFAOYSA-N

Gene Information

rat ... Ace(24310)

General description

6-Mercaptohexanoic acid (MHA) forms a self-assembled monolayer (SAM) that is used to cap a variety of materials like gold nanoparticles (AuNPs) and indium phosphide quantum dots (InPQDs). It is majorly used in immobilizing the surface molecules, which enhance the physio-chemical and electrical properties.[1][2][3]

Application

MHA can be used in the surface modification of InP quantum nanowires for their potential uage in fluorescent chemical sensors that monitor cyanide ions.[4] MHA may also be covalently bonded with liposomes that can form a SAM on the surface of gold for use as an insect odorant receptor in biosensor based applications.[5]
Used to prepare thiol-functionalized 1.5 nm gold nanoparticles via a ligand exchange process with triphenylphosphine-stabilized nanoparticles.[6] This compound is used in self-assembly to produce hydrophilic SAMs. The resulting monolayers which are terminated with carboxylic acids can be further functionalized[7] with various amines and alcohols to introduce more complex end groups. Can also be used to control the rate of aggregation of gold colloidal nanoparticles using pH.[8]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wenjuan Wei et al.
Journal of microbiology, immunology, and infection = Wei mian yu gan ran za zhi, 50(6), 821-830 (2016-01-05)
Treatment of Acinetobacter baumannii infections is challenging owing to widespread multidrug-resistant A. baumannii (MDR-AB) and the lack of novel agents. Although recent data suggest that levofloxacin (LVX) may have unique activity against MDR-AB in combination with colistin (CST), further preclinical
An Ultrasensitive Electrochemical Impedance-Based Biosensor Using Insect Odorant Receptors to Detect Odorants
Khadka R, et al.
Biosensors And Bioelectronics, 183, 441-445 (2018)
Panagiotis Papadopoulos et al.
Food microbiology, 84, 103249-103249 (2019-08-20)
Recently, there has been an increased tendency towards raw-milk consumption, which may pose a consumer risk, due to the possible presence of human pathogenic microorganisms, such as Staphylococcus aureus and even methicillin-resistant S. aureus (MRSA). The prevalence of S. aureus
Fredrik Boulund et al.
BMC genomics, 18(1), 682-682 (2017-09-04)
Fluoroquinolones are broad-spectrum antibiotics used to prevent and treat a wide range of bacterial infections. Plasmid-mediated qnr genes provide resistance to fluoroquinolones in many bacterial species and are increasingly encountered in clinical settings. Over the last decade, several families of
Electrochemical genoassays on gold-coated magnetic nanoparticles to quantify genetically modified organisms (GMOs) in food and feed as GMO percentage
Placido A, et al.
Biosensors And Bioelectronics, 110, 147-154 (2018)

Articles

Recent research highlights tunable properties of inorganic nanoparticles, driving interest in optoelectronics.

Recent research highlights tunable properties of inorganic nanoparticles, driving interest in optoelectronics.

Recent research highlights tunable properties of inorganic nanoparticles, driving interest in optoelectronics.

Recent research highlights tunable properties of inorganic nanoparticles, driving interest in optoelectronics.

See All

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service