Skip to Content
Merck
All Photos(4)

Key Documents

674613

Sigma-Aldrich

Tetrakis(dimethylamino)ethylene

Synonym(s):

Octamethylethylenetetramine, TDAE

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)2N]2C=C[N(CH3)2]2
CAS Number:
Molecular Weight:
200.32
Beilstein:
1705900
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

Assay

≥85.0%

Quality Level

form

liquid

reaction suitability

reaction type: C-C Bond Formation

refractive index

n/D 1.48 ((lit.))
n20/D 1.48 (lit.)

bp

59 °C ((lit.))
59 °C/0.9 mmHg (lit.)

density

0.861 g/mL at 25 °C ((lit.))
0.861 g/mL at 25 °C (lit.)

functional group

amine

SMILES string

[CN(C)\C(N(C)C)=C(\N(C)C)N(C)C]
CN(C)\C(N(C)C)=C(\N(C)C)N(C)C

InChI

[1S/C10H24N4/c1-11(2)9(12(3)4)10(13(5)6)14(7)8/h1-8H3]
1S/C10H24N4/c1-11(2)9(12(3)4)10(13(5)6)14(7)8/h1-8H3

InChI key

[CBXRMKZFYQISIV-UHFFFAOYSA-N]
CBXRMKZFYQISIV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Involved in TDAE methodology for coupling and intramolecular Buchwald reactions

Acts as a reducing agent

Reactant involved in:
  • Dioxygenation
  • Insertion of aluminum anion into the C-N bond
  • Chemiluminescent reactions with oxygen
Reagent used to convert α-bromoketones into 1,4-diketones.

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

127.4 °F - closed cup

Flash Point(C)

53 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wen-Bin Luo et al.
Advanced materials (Deerfield Beach, Fla.), 27(43), 6862-6869 (2015-09-25)
Porous AgPd-Pd composite nanotubes (NTs) are used as an efficient bifunctional catalyst for the oxygen reduction and evolution reactions in lithium-oxygen batteries. The porous NT structure can facilitate rapid O2 and electrolyte diffusion through the NTs and provide abundant catalytic
Patrick Weis et al.
Physical chemistry chemical physics : PCCP, 21(35), 18877-18892 (2019-08-23)
We present high-resolution trapped ion mobility spectrometry (TIMS) measurements for fullerene ions in molecular nitrogen. Three different charge states were studied (monocations, monoanions and dianions) with fullerenes ranging in size from C60 to C150. Ions were prepared by either electrospray
Tetrahedron Letters, 47, 5565-5565 (2006)
Kyungwho Choi et al.
ACS applied materials & interfaces, 10(28), 23891-23899 (2018-06-28)
Thermoelectric (TE) energy conversion with nontraditional organic materials is promising in wearable electronics and roll-to-roll manufacturing because of mechanical flexibility, lightweight, and easy processing. Although typical organic materials have a benefit of low thermal conductivity that creates a large temperature
Hong Wang et al.
Advanced materials (Deerfield Beach, Fla.), 27(43), 6855-6861 (2015-10-02)
Hybrids of carbon nanotubes (CNTs) and poly(3,4-ethylenedioxythiophene) (PEDOT) treated by tetrakis(dimethylamino)ethylene (TDAE) have large n-type voltages in response to temperature differences. The reduced carrier concentration by TDAE reduction and partially percolated CNT networks embedded in the PEDOT matrix result in

Articles

In 2001, Professor William Dolbier, Jr., at the University of Florida reported1 an approach to nucleophilic trifluoromethylation based on the generation of a trifluoromethyl anion using CF3I in the presence of a powerful twoelectron reductant, tetrakis(dimethylamino)ethylene (TDAE)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service