Skip to Content
Merck
All Photos(1)

Key Documents

776734

Sigma-Aldrich

4-(2,3-Dihydro-1,3-dimethyl-1H-benzimidazol-2-yl)-N,N-dimethylbenzenamine

97% (HPLC)

Synonym(s):

4-(1,3-Dimethyl-2,3-dihydro-1H-benzoimidazol-2-yl)phenyl)dimethylamine, N-DMBI

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H21N3
CAS Number:
Molecular Weight:
267.37
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

97% (HPLC)

form

solid

mp

105-110 °C

SMILES string

CN(C)c1ccc(cc1)C2N(C)c3ccccc3N2C

InChI

1S/C17H21N3/c1-18(2)14-11-9-13(10-12-14)17-19(3)15-7-5-6-8-16(15)20(17)4/h5-12,17H,1-4H3

InChI key

AKIIMLCQTGCWQQ-UHFFFAOYSA-N

General description

4-(2,3-Dihydro-1,3-dimethyl-1H-benzimidazol-2-yl)-N,N-dimethylbenzenamine is a semiconducting organic molecule with a π-conjugated polycyclic system. It is a strong electron donor molecule that can be used for n-type doping. It shows conductivity of ~2 × 10−3 S/cm as a dopant. It also acts as a reagent for the reductive transformation of organic compounds.

Application

Air stable n-type dopant for n-channel Organic Thin Film Transistors (OTFTs) and solar cells (OPVs).
It is mainly used as a semiconductor based polymer for the fabrication of electronic devices, which include organic thin film transistors (OTFTs), polymeric solar cells (PSCs) and organic light emitting diodes (OLEDs).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cho, Namchul;
Advanced Engineering Materials, 1, 1148-1153 (2011)
2-(2-Methoxyphenyl)-1, 3-dimethyl-1 H-benzoimidazol-3-ium iodide as a new air-stable n-type dopant for vacuum-processed organic semiconductor thin films
Wei P, et al.
Journal of the American Chemical Society, 134(9), 3999-4002 (2012)
Bhawani Chamlagain et al.
Food science & nutrition, 6(1), 67-76 (2018-02-02)
The
Molecular orbital analysis of selected organic p-type and n-type conducting small molecules
Cagardova D and Vladimir L
Acta Chimica Slovaca, 10(1), 6-16 (2017)
Himanshu Shekhar et al.
Scientific reports, 10(1), 7594-7594 (2020-05-07)
Organic photodiodes (OPDs) for its interesting optoelectronic properties has the potential to be utilized with complementary metal-oxide-semiconductor (CMOS) circuit for imaging, automotive, and security based applications. To achieve such a hybrid device as an image sensor, it is imperative that

Articles

Fabrication procedure of organic field effect transistor device using a soluble pentacene precursor.

Solution-processed organic photovoltaic devices (OPVs) have emerged as a promising clean energy generating technology due to their ease of fabrication, potential to enable low-cost manufacturing via printing or coating techniques, and ability to be incorporated onto light weight, flexible substrates.

Solution-processed organic photovoltaic devices (OPVs) have emerged as a promising clean energy generating technology due to their ease of fabrication, potential to enable low-cost manufacturing via printing or coating techniques, and ability to be incorporated onto light weight, flexible substrates.

Solution-processed organic photovoltaic devices (OPVs) have emerged as a promising clean energy generating technology due to their ease of fabrication, potential to enable low-cost manufacturing via printing or coating techniques, and ability to be incorporated onto light weight, flexible substrates.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service