Skip to Content
Merck
All Photos(1)

Key Documents

674591

Sigma-Aldrich

(R)-(+)-3,3′-Bis(3,5-bis(trifluoromethyl)phenyl)-1,1′-bi-2-naphthol

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H18F12O2
CAS Number:
Molecular Weight:
710.51
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

form

solid

optical activity

[α]22/D 45°, c = 1 in chloroform

mp

216-220 °C

functional group

fluoro

SMILES string

FC(F)(F)c1cc(cc(c1)c2cc3c(c(c2O)c4c5c(cc(c4O)c6cc(cc(c6)C(F)(F)F)C(F)(F)F)cccc5)cccc3)C(F)(F)F

InChI

1S/C36H18F12O2/c37-33(38,39)21-9-19(10-22(15-21)34(40,41)42)27-13-17-5-1-3-7-25(17)29(31(27)49)30-26-8-4-2-6-18(26)14-28(32(30)50)20-11-23(35(43,44)45)16-24(12-20)36(46,47)48/h1-16,49-50H

InChI key

PGXMYJSTCAQJBY-UHFFFAOYSA-N

Application

Precursor to a chiral Bronsted acid (680184) used to catalyze an enantioselective aza Diels-Alder reaction providing bicyclic lactams. Rare earth metal complexes of this chiral binapthol catalyze an intramolecular hydroamination of amino olefins leading to chiral pyrrolidines.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hua Liu et al.
Organic letters, 8(26), 6023-6026 (2006-12-15)
[Structure: see text] The first chiral Brønsted acid-catalyzed asymmetric direct aza hetero-Diels-Alder reaction has been described. The phosphoric acids, prepared from binol and H8-binol derivatives, have shown catalytic ability for the reaction of cyclohexenone with N-PMP-benzaldimine. A chiral phosphoric acid
Denis V Gribkov et al.
Journal of the American Chemical Society, 128(11), 3748-3759 (2006-03-16)
Chiral 3,3'-bis(trisarylsilyl)-substituted binaphtholate rare earth metal complexes (R)-[Ln{Binol-SiAr3}(o-C6H4CH2NMe2)(Me2NCH2Ph)] (Ln = Sc, Lu, Y; Binol-SiAr3 = 3,3'-bis(trisarylsilyl)-2,2'-dihydroxy-1,1'-binaphthyl; Ar = Ph (2-Ln), 3,5-xylyl (3-Ln)) and (R)-[La{Binol-Si(3,5-xylyl)3}{E(SiMe3)2}(THF)2] (E = CH (4a), N (4b)) are accessible via facile arene, alkane, and amine elimination. They

Articles

We present an article concerning BINOL and Derivatives.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service