Skip to Content
Merck
All Photos(2)

Key Documents

595721

Sigma-Aldrich

(R)-(+)-3,3′-Dibromo-1,1′-bi-2-naphthol

97%

Synonym(s):

(R)-3,3′-Dibromo-1,1′-bi-2-naphthol, (R)-3,3′-Dibromo-[1,1′-Binaphthalene]-2,2′-diol, (R)-Dibromo-1,1′-Bi-2,2′-naphthol, (R)-Dibromo-1,1′-Binaphthalene-2,2′-diol, (R)-Dibromo-1,1-Binaphthol, (R)-Dibromo-BINOL, (R)-Dibromo-bi-2-naphthol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H12Br2O2
CAS Number:
Molecular Weight:
444.12
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

mp

256-260 °C

functional group

bromo

InChI

1S/C20H12Br2O2/c21-15-9-11-5-1-3-7-13(11)17(19(15)23)18-14-8-4-2-6-12(14)10-16(22)20(18)24/h1-10,23-24H

InChI key

BRTBEAXHUYEXSY-UHFFFAOYSA-N

Application

(R)-(+)-3,3′-Dibromo-1,1′-bi-2-naphthol reacts with zirconium(IV) tert-butoxide to form a chiral zirconium complex, which can efficiently catalyze:
  • anti-selective catalytic asymmetric aldol reactions of silyl enol ethers with aldehydes
  • asymmetric intramolecular [3+2] cycloaddition of hydrazone/olefins

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Asymmetric intramolecular [3+2] cycloaddition reactions of acylhydrazones/olefins using a chiral zirconium catalyst.
Kobayashi S, et al.
Journal of the American Chemical Society, 124(46), 13678-13679 (2002)
Highly anti-selective catalytic asymmetric aldol reactions.
Ishitani H, et al.
Journal of the American Chemical Society, 122(22), 5403-5404 (2000)

Articles

We present an article concerning BINOL and Derivatives.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service