Skip to Content
Merck
All Photos(1)

Key Documents

674044

Sigma-Aldrich

Poly(methacrylic acid, sodium salt) solution

average Mw 4,000-6,000, 40 wt. % in H2O

Synonym(s):

Sodium polymethacrylate, Sodium polymethacrylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH2C(CH3)(CO2Na)]n
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

mol wt

average Mw 4,000-6,000

Quality Level

concentration

40 wt. % in H2O

refractive index

n20/D 1.430

density

1.251 g/mL at 25 °C

SMILES string

CC(=C)C(=O)O[Na]

InChI

1S/C4H6O2.Na/c1-3(2)4(5)6;/h1H2,2H3,(H,5,6);/q;+1/p-1

InChI key

SONHXMAHPHADTF-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T S Anirudhan et al.
Journal of environmental radioactivity, 116, 141-147 (2012-11-17)
The use of a carboxylate-functionalized graft copolymer (PGTDC-COOH) based on titanium dioxide-densified cellulose (TDC) for the removal and recovery of thorium(IV) [Th(IV)] from industrial wastewater is reported in this paper. Infrared spectroscopy and X-ray diffractometer were used to characterize the
Jeremy Lee et al.
Journal of chromatography. A, 1270, 162-170 (2012-11-28)
We introduce a chromatography method for purification of large proteins and viruses that works by capturing them at a non-reactive hydrophilic surface by their mutual steric exclusion of polyethylene glycol (PEG). No direct chemical interaction between the surface and the
Ye-Zi You et al.
Methods in molecular biology (Clifton, N.J.), 948, 121-132 (2012-10-17)
Bioreducible polycations, which possess disulfide linkages in the backbone, have appeared as promising gene delivery carriers due to their high stability in extracellular physiological condition and bioreduction-triggered release of genetic materials, as well as reduced cytotoxicity because intracellular cytosol is
Qiangying Yi et al.
Langmuir : the ACS journal of surfaces and colloids, 28(29), 10822-10829 (2012-06-27)
Microcapsules composed of weak polyelectrolytes modified with UV-responsive benzophenone (BP) groups were fabricated by the layer-by-layer (LbL) technique. Being exposed to UV lights, capsules shrunk in the time course of minutes at irradiation intensity of 5 mW/cm(2). The shrinkage adjusted
Loredana Ricciardi et al.
Dalton transactions (Cambridge, England : 2003), 41(36), 10923-10925 (2012-08-14)
The grafting of a 2-picolylamine Pt(II) complex into polymethacrylic acid has been successfully performed. The obtained polymer is water soluble, and it represents the first example of a platinum-containing polymer able to photogenerate singlet oxygen.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service