Skip to Content
Merck
All Photos(1)

Documents

416029

Sigma-Aldrich

Poly(acrylic acid, sodium salt) solution

average Mw ~8,000, 45 wt. % in H2O

Synonym(s):

PAA, Sodium polyacrylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH2CH(CO2Na)]n
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

viscous liquid

mol wt

average Mw ~8,000

concentration

45 wt. % in H2O

refractive index

n20/D 1.428

density

1.3 g/mL at 25 °C

SMILES string

[Na]OC(=O)C=C

InChI

1S/C3H4O2.Na/c1-2-3(4)5;/h2H,1H2,(H,4,5);/q;+1/p-1

InChI key

NNMHYFLPFNGQFZ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

  • Poly(acrylic acid) Hydrogel for Mucoadhesives: This study focuses on the radiation crosslinking of poly(acrylic acid) to create a bioadhesive hydrogel, which can be crucial for drug delivery applications, especially in mucoadhesive drug delivery systems (YC Nho, JS Park, YM Lim, 2014).
  • Hydrogel for Dye Removal: Development of a poly(acrylic acid) based hydrogel with fast adsorption rates and high capacity for removing cationic dyes, relevant for environmental cleanup and potentially for purifying biological fluids in pharmaceutical processes (Z Yuan, J Wang, Y Wang, Q Liu, Y Zhong, Y Wang, 2019).
  • Composite Hydrogels for Swelling Properties: Investigation into poly(acrylic acid-co-acrylamide) composite hydrogels, studying their swelling properties which are essential for understanding the behavior of hydrogels used in drug delivery systems and tissue engineering (WM Cheng, XM Hu, YY Zhao, MY Wu, ZX Hu, XT Yu, 2017).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Imran Khimji et al.
Chemical communications (Cambridge, England), 49(13), 1306-1308 (2013-01-11)
The melting temperature of duplex DNA is much higher in polyanions than in non-ionic polymers with similar ionic strength, suggesting an additional electrostatic contribution on top of the excluded volume effect.
Chunjiao Zhou et al.
Journal of nanoscience and nanotechnology, 13(7), 4627-4633 (2013-08-02)
Poly(acrylic acid) (PAA) coated-Fe3O4 superparamagnetic nano-composites were synthesized through a solvothermal technique by using cheap and environmental friendly iron salts and PAA. Each nano-composite was composed of many small primary nanocrystals. The as-synthesized products were characterized by X-ray diffraction (XRD)
Lei Liu et al.
ACS nano, 7(2), 1368-1378 (2013-01-04)
Hierarchical FeOOH nanostructure array films constructed by different nanosized building blocks can be synthesized at the air-water interface via a bio-inspired gas-liquid diffusion method. In this approach, poly(acrylic acid) (PAA) as a crystal growth modifier plays a crucial role in
Daniel L Miller et al.
The Annals of thoracic surgery, 95(3), 1050-1056 (2013-01-22)
Skeletal chest wall reconstruction can be a challenge, depending on the indication, location, and health of the patient; various materials are available. Recently, biomaterials that are remodelable (bovine pericardium patch; Veritas, Synovis Life Technologies Inc, St Paul, MN) or absorbable
Michael Dietzsch et al.
Langmuir : the ACS journal of surfaces and colloids, 29(9), 3080-3088 (2013-02-08)
Scale formation, the deposition of certain minerals such as CaCO3, MgCO3, and CaSO4·2H2O in industrial facilities and household devices, leads to reduced efficiency or severe damage. Therefore, incrustation is a major problem in everyday life. In recent years, double hydrophilic

Articles

Laboratory safety tools: Offers educational tools for chemistry, prioritizing customer health and safety, with online support available.

Laboratory safety tools: Offers educational tools for chemistry, prioritizing customer health and safety, with online support available.

Laboratory safety tools: Offers educational tools for chemistry, prioritizing customer health and safety, with online support available.

Laboratory safety tools: Offers educational tools for chemistry, prioritizing customer health and safety, with online support available.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service