Skip to Content
Merck
All Photos(1)

Key Documents

65800

Sigma-Aldrich

2-Methylanthraquinone

technical, ≥95% (HPLC)

Synonym(s):

2-MAQ

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H10O2
CAS Number:
Molecular Weight:
222.24
Beilstein:
2050523
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Assay:
≥95% (HPLC)

grade

technical

Quality Level

Assay

≥95% (HPLC)

impurities

3-4% 1-methylanthraquinone

bp

236-238 °C/10 mmHg (lit.)

mp

170-173 °C (lit.)

functional group

ketone

SMILES string

Cc1ccc2C(=O)c3ccccc3C(=O)c2c1

InChI

1S/C15H10O2/c1-9-6-7-12-13(8-9)15(17)11-5-3-2-4-10(11)14(12)16/h2-8H,1H3

InChI key

NJWGQARXZDRHCD-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

2-Methylanthraquinone may be used in the preparation of potential bioreducible anthraquinone derivatives.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

408.2 °F - closed cup

Flash Point(C)

209 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

D Biswas et al.
Bioresource technology, 102(2), 1284-1288 (2010-09-11)
Projected decline in future wood resources has prompted researchers to try various additives in existing pulping processes for fiber yield improvement. Many studies have been conducted in the past aimed at improving kraft pulp yield with the use of additives
Giampaolo Gori et al.
The Annals of occupational hygiene, 53(1), 27-32 (2008-11-04)
A new gas chromatographic/mass spectrometric (GC/MS) method was developed to detect 2-methylanthraquinone (2-MeA) in wood dust. 2-MeA is present in teak wood (a suspected human carcinogen) but not in oak, beech, mahogany, birch, ash or pine. The method involved collection
Emily L Whitson et al.
Journal of natural products, 75(3), 394-399 (2012-02-09)
Barleria alluaudii and Diospyros maritima were both investigated as part of an ongoing search for synergistic TRAIL (tumor necrosis factor-α-related apoptosis-inducing ligand) sensitizers. As a result of this study, two naphthoquinone epoxides, 2,3-epoxy-2,3-dihydrolapachol (1) and 2,3-epoxy-2,3-dihydro-8-hydroxylapachol (2), both not previously
2-Methylanthraquinone derivatives as potential bioreductive alkylating agents.
Lin TS, et al.
Journal of Medicinal Chemistry, 23(11), 1237-1242 (1980)
S Li et al.
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica, 16(11), 675-676 (1991-11-01)
This paper reports the identification of four compounds isolated from the cortex of Morinda officinalis growing in Guangdong Province. These compounds are beta-sitosterol (I), 2-methyl-anthraquinone(II), rubiadin-1-methyl ether(III) and 24-ethylcholesterol(VI). Compounds II and VI are isolated from Morinda Genus for the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service