Skip to Content
Merck
All Photos(1)

Documents

655821

Sigma-Aldrich

2-Acrylamido-2-methyl-1-propanesulfonic acid sodium salt solution

50 wt. % in H2O

Synonym(s):

2-Methyl-2-[(1-oxo-2-propenyl)amino]-1-propanesulfonic acid sodium, Sodium 2-acrylamido-2-methyl-1-propanesulfonate, Sodium acryloyldimethyltaurate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=CHCONHC(CH3)2CH2SO3Na
CAS Number:
Molecular Weight:
229.23
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

concentration

50 wt. % in H2O

refractive index

n20/D 1.4220 (lit.)

density

1.2055 g/mL at 25 °C (lit.)

SMILES string

[Na+].CC(C)(CS([O-])(=O)=O)NC(=O)C=C

InChI

1S/C7H13NO4S.Na/c1-4-6(9)8-7(2,3)5-13(10,11)12;/h4H,1,5H2,2-3H3,(H,8,9)(H,10,11,12);/q;+1/p-1

InChI key

FWFUWXVFYKCSQA-UHFFFAOYSA-M

General description

2-Acrylamido-2-methyl-1-propanesulfonic acid sodium salt (AMPS Na) is a monomer that belongs to the class of sulfonic acid monomers. It is commonly used in the production of various polymers, such as polyacrylamide co-polymers, poly(AMPS), AMPS-functionalized polymers, and hydrogels . These polymers are used in paints, adhesives, textiles, paper manufacturing, personal care products, coating, and adhesive applications. Additionally, AMPSNa also serves as a comonomer in combination with other acrylic monomers to enhance the scrub resistance and dispersant performance of paper coatings and paint emulsions. It can be used as a dopant and a protonating agent for conducting polymers. It is used in a variety of electronic applications.

Application

2-Acrylamido-2-methyl-1-propanesulfonic acid sodium salt (AMPS Na) can be used:
  • As a monomer in the formation of polyelectrolyte copolymer gels for potential application in bioengineering, biomedicine, and water purification .
  • In the fabrication of Schottky diodes, humidity sensors, and lithium-ion batteries.
  • As a monomer in the synthesis of a hydrogel nanocomposite applicable as a potential adsorbent for dyes .

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aurora Mejia et al.
ACS omega, 4(6), 11119-11125 (2019-08-29)
High molar masses homopolymers of both acrylamide (AM) and 2-acrylamido-2-methylpropanesulfonate (AMPS) as well as poly(AM-stat-AMPS) exhibiting a large range copolymer composition has been obtained via the optimization of a purely adiabatic gel process. Monomer concentrations ranging from 2.0 to 3.47
Fabrication of Schottky diode based on Zn electrode and polyaniline doped with 2-acrylamido-2-methyl-1-propanesulfonic acid sodium salt
Ebrahim SM
Journal of Polymer Research, 16(5), 481-487 (2009)
Polymeric absorbent for water sorption based on chemically crosslinked poly (acrylamide/2-acrylamido-2-methyl-1-propanesulfonic acid sodium salt) hydrogels
Uzum OB, et al.
Polymer Bull., 57(5), 703-712 (2006)
Cyclic voltammetry and impedance studies of electrodeposited polypyrrole nanoparticles doped with 2-acrylamido-2-methyl-1-propanesulfonic acid sodium salt
Ebrahim SM, et al.
Thin Solid Films, 518(15), 4100-4105 (2010)
Ranjith Krishna Pai et al.
Journal of the American Chemical Society, 130(39), 13074-13078 (2008-09-05)
In the biomineralization process, the changes in conformation of organic matrix may be a widespread phenomenon. Investigation of the structural relationship between organic and inorganic materials is the main subject. The approach taken was to extract quantitative information of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service