Skip to Content
Merck
All Photos(1)

Key Documents

645605

Sigma-Aldrich

Trimethyl(methylcyclopentadienyl)platinum(IV)

98%

Synonym(s):

MeCpPtMe3

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C5H4CH3Pt(CH3)3
CAS Number:
Molecular Weight:
319.30
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Assay

98%

form

low-melting solid

reaction suitability

core: platinum

mp

30-31 °C (lit.)

storage temp.

2-8°C

SMILES string

C[Pt](C)C.C[C]1[CH][CH][CH][CH]1

InChI

1S/C6H7.3CH3.Pt/c1-6-4-2-3-5-6;;;;/h2,4H,3H2,1H3;3*1H3;

InChI key

AGQKROMSWCHOND-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Trimethyl(methylcyclopentadienyl)platinum(IV) is a crystalline organometallic precursor for platinum and is used in microelectronics, catalysis, photonics, and chemical sensing processes. It is also utilized as a precursor for preparing platinum nanoparticles, tuning particle size for better activity and stability of the platinum catalyst.

Application

Trimethyl(methylcyclopentadienyl)platinum(IV) can be used:
  • As a precursor to deposit Pt on tungsten oxide surface by atomic layer deposition method. This acts as a hybrid anode catalyst for PEM fuel cells.
  • To fabricate high performance and stable Si photocathode for solar water splitting reaction.
  • As a precursor to deposit Pt nanoparticles on TiO2 nanotube films for highly efficient gas sensors. Pt nanoparticles significantly enhances the sensitivity.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Oral - Acute Tox. 2 Dermal - Aquatic Acute 1 - Skin Sens. 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

131.0 °F - closed cup

Flash Point(C)

55 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Electron induced surface reactions of the organometallic precursor trimethyl (methylcyclopentadienyl) platinum (IV).
The Journal of Physical Chemistry C, 113(6), 2487-2496 (2009)
Nanostructured Platinum Catalysts by Atomic-Layer Deposition for Solid-Oxide Fuel Cells
Advanced Energy Materials, 2(6), 651-654 (2012)

Articles

Spintronics offer breakthroughs over conventional memory/logic devices with lower power, leakage, saturation, and complexity.

Spintronics offer breakthroughs over conventional memory/logic devices with lower power, leakage, saturation, and complexity.

Spintronics offer breakthroughs over conventional memory/logic devices with lower power, leakage, saturation, and complexity.

Spintronics offer breakthroughs over conventional memory/logic devices with lower power, leakage, saturation, and complexity.

See All

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service