Skip to Content
Merck
All Photos(1)

Key Documents

110248

Sigma-Aldrich

Amino-2-propanol

93%

Synonym(s):

(±)-1-Amino-2-propanol, (±)-Isopropanolamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(OH)CH2NH2
CAS Number:
Molecular Weight:
75.11
Beilstein:
605275
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

vapor density

2.6 (vs air)

Quality Level

vapor pressure

<1 mmHg ( 20 °C)

Assay

93%

impurities

7% 2-amino-1-propanol

refractive index

n20/D 1.4478 (lit.)

bp

160 °C (lit.)

mp

−2 °C (lit.)

density

0.973 g/mL at 25 °C (lit.)

SMILES string

CC(O)CN

InChI

1S/C3H9NO/c1-3(5)2-4/h3,5H,2,4H2,1H3

InChI key

HXKKHQJGJAFBHI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Amino-2-propanol is used to study the effect of temperature and water content on the molecular structure and hydrogen bonding of amino-2-propanol, which has been examined by Fourier transform near-infrared (FT-NIR) spectroscopy[1].

Application

Amino-2-propanol can be used as a reactant to prepare:
  • 5-methyl-2-oxazolidinone by oxidative carbonylation reaction using Pd(OAc)2/I2 catalyst.[2]
  • Bio-based polyurethanes by reacting with epoxidized soybean oil via ring-opening and amidation reaction.[3]

It can also be used as a solvent as well as a template to synthesize layered aluminophosphate containing a racemic mixture of isopropanolamine.[4]

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

159.8 °F - closed cup

Flash Point(C)

71 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of bio-based polyurethanes from epoxidized soybean oil and isopropanolamine
Miao S, et al.
Journal of Applied Polymer Science, 127(3), 1929-1936 (2013)
Dual function of racemic isopropanolamine as solvent and as template for the synthesis of a new layered aluminophosphate:[NH3CH2CH (OH) CH3] 3{\textperiodcentered} Al3P4O16
Y Hong-Ming, et al.
Journal of Solid State Chemistry, 151(1), 145-149 (2000)
An Efficient Synthesis of 2-Oxazolidinones by Palladium-catalyzed Oxidative Carbonylation of ?-Aminoalcohols and 2-Aminophenol under Mild Conditions
Fu-Wei Li, et al.
Chin. J. Chem., 23(5), 643-645 (2005)
Krzysztof Zdzisław Haufa et al.
Applied spectroscopy, 64(3), 351-358 (2010-03-13)
The effect of temperature and water content on the molecular structure and hydrogen bonding of 2-aminoethanol (2AE), 1-amino-2-propanol (2AP), and 3-amino-1-propanol (3AP) has been examined by Fourier transform near-infrared (FT-NIR) spectroscopy. The experimental spectra were analyzed using the two-dimensional (2D)
Steven B Hawthorne et al.
Environmental science & technology, 39(10), 3639-3645 (2005-06-15)
Soil and groundwater samples were collected at the site of a former chemical processing plant in areas impacted by accidental releases of MEA (monoethanolamine) and IPA (2-propanolamine or isopropanolamine). Although their use had ceased ca. 10 years before sample collection

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service