Skip to Content
Merck
All Photos(1)

Key Documents

642452

Sigma-Aldrich

Molybdenum(V) chloride

anhydrous, powder, 99.99% trace metals basis (excluding W)

Synonym(s):

Molybdenum pentachloride, Molybdenum(5+) chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
MoCl5
CAS Number:
Molecular Weight:
273.21
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
99.99% trace metals basis (excluding W)
grade:
anhydrous
form:
powder

grade

anhydrous

Quality Level

vapor pressure

1.75 mmHg ( 25 °C)
131 mmHg ( 250 °C)

Assay

99.99% trace metals basis (excluding W)

form

powder

impurities

≤150.0 ppm Trace Metal Analysis

bp

268 °C (lit.)

mp

194 °C (lit.)

density

2.928 g/mL at 25 °C (lit.)

application(s)

battery manufacturing

SMILES string

Cl[Mo](Cl)(Cl)(Cl)Cl

InChI

1S/5ClH.Mo/h5*1H;/q;;;;;+5/p-5

InChI key

GICWIDZXWJGTCI-UHFFFAOYSA-I

Looking for similar products? Visit Product Comparison Guide

General description

Molybdenum(V) chloride, also called as molybdenum pentachloride is a Lewis acid with high oxidation potential. It is widely used as a catalyst in the field of organic synthesis and polymer chemistry. It is also used as a chemical vapor deposition precursor.

Application

Molybdenum(V) chloride can be used:
  • As a catalyst for amidation of secondary benzyl alcohols.
  • As a precursor to fabricate MoS2 thin films by atomic layer deposition method.
  • As a primary catalyst for coordination polymerization of butadiene.
  • To fabricate superior anode materials for Na-ion and Li-ion batteries.
  • As a dual-function redox mediator for Li–O2 batteries to overcome thehigh polarization and low energy density issues.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

MoCl5 as a dual-function redox mediator for Li-O2 batteries
Xin-Gai Wang, et al.
Journal of Materials Chemistry, 7, 14239-14243 (2019)
m-Cresol-substituted triethyl aluminum/MoCl5/TBP catalytic system for coordination polymerization of butadiene
Peipei Li, et al.
Iranian Polymer Journal, 27, 537-544 (2018)
Molybdenum(V) chloride-catalyzed amidation of secondary benzyl alcohols with sulfonamides and carbamates
Ch. Raji Reddy, et al.},
Tetrahedron Letters, 48, 7169-7172 (2007)
Atomic layer deposition of a MoS2 film
Lee Kheng Tan, et al.},
Nanoscale, 6, 10584-10588 (2014)
Towards high-volumetric performance of Na/Li-ion batteries: A better anode material with molybdenum pentachloride--graphite intercalation compounds (MoCl 5--GICs
Zheng Li, et al.
Journal of Materials Chemistry, 8, 2430-2438 (2020)

Articles

The diversity of applications and nanostructured materials accessible using ultrasonic spray methods are highlighted in this article.

The diversity of applications and nanostructured materials accessible using ultrasonic spray methods are highlighted in this article.

The diversity of applications and nanostructured materials accessible using ultrasonic spray methods are highlighted in this article.

The diversity of applications and nanostructured materials accessible using ultrasonic spray methods are highlighted in this article.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service