Skip to Content
Merck
All Photos(2)

Documents

263974

Sigma-Aldrich

Tungsten(IV) chloride

95%

Synonym(s):

Tungsten tetrachloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
WCl4
CAS Number:
Molecular Weight:
325.65
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Assay

95%

form

powder

reaction suitability

reagent type: catalyst
core: tungsten

SMILES string

Cl[W](Cl)(Cl)Cl

InChI

1S/4ClH.W/h4*1H;/q;;;;+4/p-4

InChI key

YOUIDGQAIILFBW-UHFFFAOYSA-J

Application

The product may be used as one of the precursor reactant for the preparation of highly conductive tungsten diselenide nanosheets.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Solution-phase synthesis of highly conductive tungsten diselenide nanosheets
Antunez PD, et al.
Chemistry of Materials, 25.12, 2385-2387 (2013)
Yun-Hang Qiu et al.
Nanoscale, 11(17), 8538-8545 (2019-04-17)
We synthesize Au@WS2 hybrid nanobelts and investigate their third-order nonlinear responses mediated by a strong anti-Stokes effect. By using the femtosecond Z-scan technique and tuning the excitation photon energy (Eexc), we find the sign reversals of both nonlinear absorption coefficient
Cotton, F.A. et al.
Inorganic Chemistry, 36, 80-80 (1997)
Lin Sun et al.
Nanoscale, 11(30), 14237-14241 (2019-07-19)
The construction of the hierarchical nanostructures of inorganic materials with high stability has attracted great attention for photothermal therapy. In this paper, we presented the preparation of urchin-like LaWO4Cl nanostructures assembled with nanoribbons via a facile solvothermal reaction. X-ray diffraction
Hui-Quan Wang et al.
Materials science & engineering. C, Materials for biological applications, 80, 102-109 (2017-09-04)
Photothermal therapy (PTT) has drawn tremendous attention because of its high therapeutic efficiency in targeting cells while minimizing the damage to normal tissues and organs. Tungsten oxide (W

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service