Skip to Content
Merck
All Photos(2)

Key Documents

637335

Sigma-Aldrich

Gadolinium(III) oxide

nanopowder, <100 nm particle size (BET), 99.8% trace metals basis

Synonym(s):

Gadolinia, Gadolinium trioxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Gd2O3
CAS Number:
Molecular Weight:
362.50
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Assay

99.8% trace metals basis

form

nanopowder

reaction suitability

reagent type: catalyst
core: gadolinium

particle size

<100 nm (BET)

mp

2330 °C (lit.)

density

7.407 g/mL at 20 °C (lit.)

SMILES string

O=[Gd]O[Gd]=O

InChI

1S/2Gd.3O

InChI key

RNYFGGMJZRTZKO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrée-Anne Guay-Bégin et al.
Langmuir : the ACS journal of surfaces and colloids, 28(1), 774-782 (2011-10-06)
The performance of nanomaterials for biomedical applications is highly dependent on the nature and the quality of surface coatings. In particular, the development of functionalized nanoparticles for magnetic resonance imaging (MRI) requires the grafting of hydrophilic, nonimmunogenic, and biocompatible polymers
Nikesh Gupta et al.
International journal of nanomedicine, 7, 5491-5500 (2012-12-13)
Mesoporous silica nanoparticles coencapsulating gadolinium oxide and horseradish peroxidase (HRP) have been synthesized in the aqueous core of sodium bis-(2-ethylhexyl) sulfosuccinate (AOT)-hexane-water reverse micelle. The average diameter of these silica particles is around 25 nm and the particles are spherical
Zijian Zhou et al.
Advanced materials (Deerfield Beach, Fla.), 24(46), 6223-6228 (2012-09-14)
Monodisperse Gd(2) O(3) -embedded iron oxide (GdIO) nanoparticles can simultaneously enhance the local magnetic field intensities of each other under an external magnetic field and result in synergistic enhancement of T(1) and T(2) effects. GdIO nanoparticles have the unique property
Eva Hemmer et al.
Acta biomaterialia, 9(1), 4734-4743 (2012-09-12)
Bioimaging is an important diagnostic tool in the investigation and visualization of biological phenomena in cells and in medicine. In this context, up-converting Gd(2)O(3):Er(3+),Yb(3+) nanostructures (nanoparticles, nanorods) have been synthesized by precipitation methods and hydrothermal synthesis. Independent of size and
Natalia Abrikossova et al.
Nanotechnology, 23(27), 275101-275101 (2012-06-19)
We have previously shown that gadolinium oxide (Gd(2)O(3)) nanoparticles are promising candidates to be used as contrast agents in magnetic resonance (MR) imaging applications. In this study, these nanoparticles were investigated in a cellular system, as possible probes for visualization

Articles

Three approaches generate white light, including LED-based down-conversion for broader applications.

Three approaches generate white light, including LED-based down-conversion for broader applications.

Three approaches generate white light, including LED-based down-conversion for broader applications.

Three approaches generate white light, including LED-based down-conversion for broader applications.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service