Skip to Content
Merck
All Photos(1)

Key Documents

753645

Sigma-Aldrich

Gold nanoparticles

50 nm diameter, OD 1, stabilized suspension in 0.1 mM PBS, reactant free

Synonym(s):

Au NP, Gold colloid

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Quality Level

form

nanoparticles
suspension

packaging

poly bottle of 100 mL
poly bottle of 25 mL

storage condition

do not freeze

concentration

~3.5E+10 particles/mL

OD

1

diameter

50 nm

λmax

531-535 nm

PDI

<0.2

storage temp.

2-8°C

SMILES string

[Au]

InChI

1S/Au

InChI key

PCHJSUWPFVWCPO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Gold nanoparticles may be prepared by reduction of metal salt precursors with reducing agents, such as by citrate reduction of HAuCl4 in water.[1]

Application

Gold nanoparticles 50 nm diameter, OD 1, stabilization suspension in 0.1 mM PBS, reactant free can be potentially used as a theranostic agent for the treatment of COVID-19.[2] It may also be used in the fabrication of fiber-optic biosensors for low cost and fast detection of SARS-CoV-2.[3]
Gold nanoparticles are used in a wide range of applications. It may be used :
  • As conductors in printable inks, [4]
  • In Photodynamic therapy, [5]
  • In drug delivery and biomarkers, [6][7]
  • As calorimetric sensor, [8]
  • In biological imaging,[9]

Legal Information

Product of CytoDiagnostics, Inc.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P-FAB: a fiber-optic biosensor device for rapid detection of COVID-19
Murugan D, et al.
Transactions of the Indian National Academy of Engineering, 10(13), 1-5 (2020)
Gold Nanoparticle Sensor for the Visual Detection of Pork Adulteration in Meatball Formulation
Ali ME, et al.
Journal of Nanomaterials (2012)
Aggregation and adhesion of gold nanoparticles in phosphate buffered saline
Du S, et al.
Journal of Nanoparticle Research, 14 null
Gang Peng et al.
Nature nanotechnology, 4(10), 669-673 (2009-10-08)
Conventional diagnostic methods for lung cancer are unsuitable for widespread screening because they are expensive and occasionally miss tumours. Gas chromatography/mass spectrometry studies have shown that several volatile organic compounds, which normally appear at levels of 1-20 ppb in healthy
Tanya Stuchinskaya et al.
Photochemical & photobiological sciences : Official journal of the European Photochemistry Association and the European Society for Photobiology, 10(5), 822-831 (2011-04-02)
A 4-component antibody-phthalocyanine-polyethylene glycol-gold nanoparticle conjugate is described for use as a potential drug for targeted photodynamic cancer therapy. Gold nanoparticles (4 nm) were stabilised with a self-assembled layer of a zinc-phthalocyanine derivative (photosensitiser) and a heterobifunctional polyethylene glycol. Anti-HER2

Articles

Steven J. Oldenburg, Ph.D. provides an overview of lateral flow diagnostic assays and discusses the use of ultra-bright reporter particles based on the unique optical properties of gold nanoshells that significantly increase the sensitivity of lateral flow immunoassays.

Steven J. Oldenburg, Ph.D. provides an overview of lateral flow diagnostic assays and discusses the use of ultra-bright reporter particles based on the unique optical properties of gold nanoshells that significantly increase the sensitivity of lateral flow immunoassays.

Sustainable energy sources with high production efficiency are crucial for meeting increasing energy demand.

Steven J. Oldenburg, Ph.D. provides an overview of lateral flow diagnostic assays and discusses the use of ultra-bright reporter particles based on the unique optical properties of gold nanoshells that significantly increase the sensitivity of lateral flow immunoassays.

See All

Questions

1–10 of 13 Questions  
  1. Are the nanoparticles capped with any agents? If so, please let me know the material used as the capping agent.

    1 answer
    1. The gold nanoparticles are capped with citrate.

      Helpful?

  2. How many grams of gold nanoparticles in the 1 ml of 50nm gold nanoparticle suspension. Kindly provide this details as i need to know the diffusion rate of it.

    1 answer
    1. This is a product of CytoDiagnostics, Inc. The 50 nm gold particles have a weight concentration of 44.5 ug/mL. This is an approximation as there is some degree of variability in particle size. See the image below to review the gold nanoparticle specification ranges.

      Helpful?

  3. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  4. Which gold nanoparticle size should I choose for my application?

    1 answer
    1. The size of gold nanoparticle to use is very depandent upon the intended application. Generally, smaller particles offer better sensitivity in applications such as immunogold labeling due to less steric hindrance and the ability to bind more gold nanoparticles to the desired target. Small gold nanoparticles are less visible than larger particles, however, which must also be taken into account.As an application example, particles with a size between 30-50nm are particularly useful for the development of rapid tests such as lateral flow assays.

      Helpful?

  5. Can the Gold nanoparticles be frozen?

    1 answer
    1. No, the Gold nanoparticles should not be frozen.  If frozen, the gold nanoparticles will irreversibly aggregate, turning the solution clear.

      Helpful?

  6. What is the shape of the gold nanoparticles?

    1 answer
    1. The majority (>95%) of the gold nanoparticles are spherical.

      Helpful?

  7. How do you attach gold nanoparticles to glass?

    1 answer
    1. To view the procedure on how to attach gold nanoparticles to glass please view information in the reference - Journal of Atomic, Molecular, and Optical PhysicsVolume 2012 (2012). Article ID 683830, 6 pagesdoi:10.1155/2012/683830

      Helpful?

  8. Do I need to wash my gold nanoparticles?

    1 answer
    1. For most applications, our gold nanoparticles can be used without any additional washing steps. If you have a sensitive application that requires additional washing the best way to do so is by either centrifugation or filtration.

      Helpful?

  9. How do you attach gold nanoparticles to peptides and peptide conjugates?

    1 answer
    1. To view information on how to attach gold nanoparticles to peptides and peptide conjugates, please view the information in the reference - Rosenthal, S. J. and Wright, D. W. (Eds.). (2005), p. 91-92. NanoBiotechnology Protocols. New Jersey. Humana Press.

      Helpful?

  10. How does the storage temperature relate to shipping conditions?

    1 answer
    1. The storage conditions that a Sigma-Aldrich catalog and label recommend for products are deliberately conservative. For many products, long-term storage at low temperatures will increase the time during which they are expected to remain in specification and therefore are labeled accordingly. Where short-term storage, shipping time frame, or exposure to conditions other than those recommended for long-term storage will not affect product quality, Sigma-Aldrich will ship at ambient temperature. The products sensitive to short-term exposure to conditions other than their recommended long-term storage are shipped on wet or dry ice. Ambient temperature shipping helps to control shipping costs for our customers. At any time, our customers can request wet- or dry-ice shipment, but the special handling is at customer expense if our product history indicates that the product is stable for regular shipment.

      Helpful?

1–10 of 13 Questions  

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service