615544
Choline chloride-1,1,2,2-d4
≥98 atom % D, 98% (CP)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
HOCD2CD2N(CH3)3Cl
CAS Number:
Molecular Weight:
143.65
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
Recommended Products
isotopic purity
≥98 atom % D
Quality Level
Assay
98% (CP)
mp
302-305 °C (dec.) (lit.)
mass shift
M+4
SMILES string
[Cl-].[2H]C([2H])(O)C([2H])([2H])[N+](C)(C)C
InChI
1S/C5H14NO.ClH/c1-6(2,3)4-5-7;/h7H,4-5H2,1-3H3;1H/q+1;/p-1/i4D2,5D2;
InChI key
SGMZJAMFUVOLNK-HGFPCDIYSA-M
Packaging
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Dimerization of a metal complex through thermally induced single-crystal-to-single-crystal transformation or mechanochemical reaction.
Jie Sun et al.
Angewandte Chemie (International ed. in English), 50(31), 7061-7064 (2011-06-24)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service