All Photos(1)
About This Item
Linear Formula:
12CO2
CAS Number:
Molecular Weight:
44.00
MDL number:
UNSPSC Code:
12142100
PubChem Substance ID:
Recommended Products
vapor density
1.52 (vs air)
description
13C-depleted
isotopic purity
99.99 atom % 12C
mp
−78.5 °C (subl.) (lit.)
mass shift
depleted
SMILES string
O=[12C]=O
InChI
1S/CO2/c2-1-3/i1+0
InChI key
CURLTUGMZLYLDI-IGMARMGPSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Packaging
All gas volumes are quoted at Standard Temperature and Pressure (STP) and not at Normal Temperature and Pressure (NTP). One STP Liter is 7.7% more gas than one NTP Liter.
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Press. Gas Liquefied gas
Storage Class Code
2A - Gases
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Kees Jan van Groenigen et al.
Science (New York, N.Y.), 344(6183), 508-509 (2014-04-26)
Soils contain the largest pool of terrestrial organic carbon (C) and are a major source of atmospheric carbon dioxide (CO2). Thus, they may play a key role in modulating climate change. Rising atmospheric CO2 is expected to stimulate plant growth
Steven C Sherwood et al.
Nature, 505(7481), 37-42 (2014-01-02)
Equilibrium climate sensitivity refers to the ultimate change in global mean temperature in response to a change in external forcing. Despite decades of research attempting to narrow uncertainties, equilibrium climate sensitivity estimates from climate models still span roughly 1.5 to
Dan Li et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(1), 191-196 (2013-12-25)
New materials capable of binding carbon dioxide are essential for addressing climate change. Here, we demonstrate that amyloids, self-assembling protein fibers, are effective for selective carbon dioxide capture. Solid-state NMR proves that amyloid fibers containing alkylamine groups reversibly bind carbon
Eoin P Cummins et al.
Cellular and molecular life sciences : CMLS, 71(5), 831-845 (2013-09-21)
The capacity of organisms to sense changes in the levels of internal and external gases and to respond accordingly is central to a range of physiologic and pathophysiologic processes. Carbon dioxide, a primary product of oxidative metabolism is one such
Derek R Lovley et al.
Current opinion in biotechnology, 24(3), 385-390 (2013-03-08)
Electricity can be an energy source for microbially catalyzed production of fuels and other organic commodities from carbon dioxide. These electrobiocommodities (E-BCs) can be produced directly via electrode-to-microbe electron transfer or indirectly with electrochemically generated electron donors such as H2
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service