Skip to Content
Merck
All Photos(1)

Key Documents

422606

Sigma-Aldrich

Carbon-12C dioxide

99.9 atom % 12C

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
12CO2
CAS Number:
Molecular Weight:
44.00
EC Number:
MDL number:
UNSPSC Code:
12352119
PubChem Substance ID:
NACRES:
NA.12
Pricing and availability is not currently available.

vapor density

1.52 (vs air)

description

13C-depleted

isotopic purity

99.9 atom % 12C

mp

−78.5 °C (subl.) (lit.)

mass shift

depleted

SMILES string

O=[12C]=O

InChI

1S/CO2/c2-1-3/i1+0

InChI key

CURLTUGMZLYLDI-IGMARMGPSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

All gas volumes are quoted at Standard Temperature and Pressure (STP) and not at Normal Temperature and Pressure (NTP). One STP Liter is 7.7% more gas than one NTP Liter.
Standard products are packaged in a 450 mL carbon steel lecture bottle with brass CGA 110/180 valve. Nominal gas pressures at 21ºC are 320 psig for 10 L and 625 psig for 25 L. Care must be taken when extracting this product from the cylinder.
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Recommended products

Stainless steel control valves Z146951, Z187224 or either of stainless steel regulators Z148504 and Z148512 is recommended.

Pictograms

Gas cylinder

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Press. Gas Liquefied gas

Storage Class Code

2A - Gases

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Derek R Lovley et al.
Current opinion in biotechnology, 24(3), 385-390 (2013-03-08)
Electricity can be an energy source for microbially catalyzed production of fuels and other organic commodities from carbon dioxide. These electrobiocommodities (E-BCs) can be produced directly via electrode-to-microbe electron transfer or indirectly with electrochemically generated electron donors such as H2
Eoin P Cummins et al.
Cellular and molecular life sciences : CMLS, 71(5), 831-845 (2013-09-21)
The capacity of organisms to sense changes in the levels of internal and external gases and to respond accordingly is central to a range of physiologic and pathophysiologic processes. Carbon dioxide, a primary product of oxidative metabolism is one such
Kees Jan van Groenigen et al.
Science (New York, N.Y.), 344(6183), 508-509 (2014-04-26)
Soils contain the largest pool of terrestrial organic carbon (C) and are a major source of atmospheric carbon dioxide (CO2). Thus, they may play a key role in modulating climate change. Rising atmospheric CO2 is expected to stimulate plant growth
L V Gatti et al.
Nature, 506(7486), 76-80 (2014-02-07)
Feedbacks between land carbon pools and climate provide one of the largest sources of uncertainty in our predictions of global climate. Estimates of the sensitivity of the terrestrial carbon budget to climate anomalies in the tropics and the identification of
Trevor F Keenan et al.
Nature, 499(7458), 324-327 (2013-07-12)
Terrestrial plants remove CO2 from the atmosphere through photosynthesis, a process that is accompanied by the loss of water vapour from leaves. The ratio of water loss to carbon gain, or water-use efficiency, is a key characteristic of ecosystem function

Protocols

Separation of Methane; Acetylene; Carbon monoxide; Water; Nitrogen; Carbon dioxide; Ethane; Ethylene

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service