Skip to Content
Merck
All Photos(1)

Key Documents

578177

Sigma-Aldrich

Polypyrrole

doped, conductivity 0.5-1.5 S/cm (pressed pellet, typical), extent of labeling: ~5 wt. % loading, coated on titanium dioxide

Synonym(s):

PPy

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Quality Level

form

solid

contains

proprietary organic sulfonic acid as dopant

extent of labeling

~5 wt. % loading

solubility

H2O: insoluble
organic solvents: insoluble

SMILES string

[nH]1cccc1

InChI

1S/C4H5N/c1-2-4-5-3-1/h1-5H

InChI key

KAESVJOAVNADME-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Process up to at least 170°C in air. Process up to at least 270°C under nitrogen.Polypyrrole (PPy) is a conductive and highly stable polymer. It may be prepared by standard electrochemical technique.[1] PPy may also be prepared by reacting β-napthalene sulfonic acid (NSA) and ammonium peroxo-disulphate in aqueous medium.[2] The charges on the surfaces can be easily modified by doping the polymer during its synthesis.[1] Solubility and conductivity measurements of PPy doped with camphor sulfonic and dodecyl benzene sulfonic acid has been reported. [3]

Application

Conducting polymer.
Conductive additive for thermoplasitics and thermosets.

Packaging

Packaged in glass bottles

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fengyu Qi et al.
Biomaterials, 34(7), 1799-1809 (2012-12-12)
Electrical stimulation (ES) applied to a conductive nerve graft holds the great potential to improve nerve regeneration and functional recovery in the treatment of lengthy nerve defects. A conductive nerve graft can be obtained by a combination of conductive nerve
Gusphyl A Justin et al.
Conference proceedings : ... Annual International Conference of the IEEE Engineering in Medicine and Biology Society. IEEE Engineering in Medicine and Biology Society. Annual Conference, 2012, 1206-1209 (2013-02-01)
An electronically-controlled drug delivery system (eDDS) for the on-demand release of anti-inflammatory, anti-microbial and analgesic agents to aid in wound healing is currently under development. The loading of several drugs into conductive polymer films and their subsequent on-demand, controlled release
Darren Svirskis et al.
Therapeutic delivery, 4(3), 307-313 (2013-02-28)
Intrinsically conducting polymers, such as polypyrrole (PPy), have been utilized for drug delivery purposes as drug release rates can be tuned by electrical stimulation. Electrical stimulation can be used to switch the redox state of PPy, subsequently changing the electrostatic
Chemical Synthesis and Morphology of b?Naphthalene Sulfonic Acid-Doped Polypyrrole Micro/Nanotube.
Akinyeye R, et al.
Fullerenes, Nanotubes, and Carbon Nanostructures, 14 (2006)
N Catherine Hogan et al.
Conference proceedings : ... Annual International Conference of the IEEE Engineering in Medicine and Biology Society. IEEE Engineering in Medicine and Biology Society. Annual Conference, 2012, 803-806 (2013-02-01)
Conducting polymers are promising materials for fabrication of microelectrode arrays for both neural stimulation and recording. Our ability to engineer the morphology and composition of polypyrrole together with its suitability as an electrically addressable tissue/cell substrate have been used to

Articles

The application of conducting polymers at the interface with biology is an exciting new trend in organic electronics research.

Self-healing soft electronic materials offer potential cost savings and reduced electronic waste.

Self-healing soft electronic materials offer potential cost savings and reduced electronic waste.

Self-healing soft electronic materials offer potential cost savings and reduced electronic waste.

See All

Questions

  1. i want to coat this polymer on cotton substrate through in-situ polymerization. but it is insoluble in water. can you help me with the process of in-situ polymerization.

    1 answer
    1. In-situ polymerization usually starts with monomer, not polymer. While 578177 is polypyrrole, not pyrrole. Here is a reference paper that might be helpful: https://pubs.acs.org/doi/10.1021/acsomega.2c00327

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service