Skip to Content
Merck
All Photos(4)

Key Documents

577030

Sigma-Aldrich

Polypyrrole

greener alternative

conductivity 10-50 S/cm (pressed pellet)

Synonym(s):

PPy

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

form

solid

contains

proprietary organic sulfonic acid as dopant

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

solubility

H2O: insoluble
organic solvents: insoluble

greener alternative category

SMILES string

[nH]1cccc1

InChI

1S/C4H5N/c1-2-4-5-3-1/h1-5H

InChI key

KAESVJOAVNADME-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Polypyrrole (PPy) is a widely used highly conductive and highly stable heterocyclic polymer. It is majorly used in electrochemical applications. It is a cost-efficient conducting polymer that has a stable oxidation and high solubility in water. It exhibits a high degree of flexibility than polyaniline and has a density that facilitates a higher capacitance per unit volume.[1][2][3][4]
PPy may be prepared by a standard electrochemical technique.[5] It may also be prepared by reacting -napthalene sulfonic acid (NSA) and ammonium peroxo-disulfate in aqueous medium.[6] The charges on the surfaces can be easily modified by doping the polymer during its synthesis.[7] Solubility and conductivity measurements of PPy doped with camphor sulfonic and dodecyl benzene sulfonic acid have been reported.[8] Lectrosensitivity and lower oxidation potential of PPy make it potentially useful for drug delivery, chemical sensors, batteries, ion selective electrodes, biosensor and biochemistry research.[9]
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Green Chemistry. This product is used in energy conversion and storage, thus has been enhanced for energy efficiency. Click here for more information.

Application

Conducting polymer.
Conductive additive for thermoplasitics and thermosets.
Conductive additive for thermoplastics and thermosets. Polypropylene composites constitute the polymer electrolyte membrane fuel cell (PEMFC). Addition of PPy to polypropylene composites alters its conductivity.[7][10]
PPy can be used for a variety of applications such as:
  • electrode materials for sensors used in electrocardiography (ECG)[11]
  • formation of electrode material for a variety of energy storage applications[12]
  • encapsulation of lithium sulfide (Li2S) as a high performance cathode material[13]

PPy is mainly used in the fabrication of a variety of electrochemical devices, which include supercapacitors,[14][15] chemical sensors,[16] dye sensitized solar cells and lithium-ion batteries.[17]

Packaging

Packaged in glass bottles

Other Notes

Stable up to at least 290°C in air

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

In-situ encapsulation of nickel nanoparticles in polypyrrole nanofibres with enhanced performance for supercapacitor
Muhamad SU, et al.
Electrochimica Acta, 249(8), 9-15 (2017)
Conducting polymers with superhydrophobic effects as anticorrosion coating
Intelligent Coatings for Corrosion Control, 409-430 (2014)
Sensors Based on Conducting Polymers for Measurement of Physiological Parameters
Teran-Jimenez O, et al.
IEEE Sensors Journal, 17(8), 2492-2497 (2017)
Conducting polymers with superhydrophobic effects as anticorrosion coating
Intelligent Coatings for Corrosion Control edited by Atul Tiwari, Lloyd Hihara, James Rawlins, 409-430 (2014)
Facile synthesis of Li 2 S-polypyrrole composite structures for high-performance Li 2 S cathodes
She ZW, et al.
Energy & Environmental Science, 7(2), 672-676 (2014)

Articles

By altering the physicochemical properties, smart or intelligent drug delivery systems can be designed to deliver therapeutic molecules on-demand. Learn more about the application of stimuli-responsive materials in drug delivery.

By altering the physicochemical properties, smart or intelligent drug delivery systems can be designed to deliver therapeutic molecules on-demand. Learn more about the application of stimuli-responsive materials in drug delivery.

By altering the physicochemical properties, smart or intelligent drug delivery systems can be designed to deliver therapeutic molecules on-demand. Learn more about the application of stimuli-responsive materials in drug delivery.

By altering the physicochemical properties, smart or intelligent drug delivery systems can be designed to deliver therapeutic molecules on-demand. Learn more about the application of stimuli-responsive materials in drug delivery.

See All

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service