574643
4-Ethoxycarbonylphenylboronic acid
≥95%
Synonym(s):
4-Boronobenzoic acid 1-ethyl ester, 4-Carboethoxybenzeneboronic acid, Ethyl 4-boronobenzoate
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Linear Formula:
C2H5OCOC6H4B(OH)2
CAS Number:
Molecular Weight:
193.99
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
≥95%
form
powder or crystals
mp
135 °C (dec.) (lit.)
functional group
ester
SMILES string
CCOC(=O)c1ccc(cc1)B(O)O
InChI
1S/C9H11BO4/c1-2-14-9(11)7-3-5-8(6-4-7)10(12)13/h3-6,12-13H,2H2,1H3
InChI key
ZLNFACCFYUFTLD-UHFFFAOYSA-N
Application
Reactant involved in:
- Oxidative hydroxylation for the preparation of phenols
- Homolytic aromatic substitution
- Cross-coupling with α-bromocarbonyl compounds
- Suzuki-coupling reaction with quinoline carboxylates
- Trifluoromethylation
- Carbometalation of ynamides
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
You Zhou et al.
The Analyst, 143(15), 3628-3634 (2018-07-04)
1-Hydroxypyrene (1-HP) is a urinary metabolite of polycyclic aromatic hydrocarbons (PAHs), and can function as a convenient biomarker for human intoxication of PAH carcinogens. The development of simple 1-HP sensors with high sensitivity and fast response is highly desirable. Herein
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service