Skip to Content
Merck
All Photos(2)

Documents

143847

Sigma-Aldrich

2,4-Dichloropyrimidine

98%

Synonym(s):

2,6-Dichloropyrimidine, 2-Chloropyrimidin-4-yl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H2Cl2N2
CAS Number:
Molecular Weight:
148.98
Beilstein:
110911
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

bp

101 °C/23 mmHg (lit.)

mp

57-61 °C (lit.)

SMILES string

Clc1ccnc(Cl)n1

InChI

1S/C4H2Cl2N2/c5-3-1-2-7-4(6)8-3/h1-2H

InChI key

BTTNYQZNBZNDOR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,4-Dichloropyrimidine is a human skin sensitizer. It undergoes effective one-pot, regioselective double Suzuki coupling reaction to yield diarylated pyrimidines.

Application

2,4-Dichloropyrimidine was used in the synthesis of medicinally important 4-aryl-5-pyrimidinylimidazoles.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaohu Deng et al.
Organic letters, 8(2), 269-272 (2006-01-18)
[reaction: see text] Starting from 2,4-dichloropyrimidine, a concise synthetic route to medicinally important 4-aryl-5-pyrimidinylimidazoles is described. Sequential substitution of the 4- and 2-chloro groups using a regioselective Sonogashira coupling, followed by nucleophilic substitution, led to pyrimidinylalkyne derivatives, which were then
Samantha C Anderson et al.
Synthesis, 2010(16), 2721-2724 (2010-08-01)
An effective one-pot, regioselective double Suzuki coupling of 2,4-dichloropyrimidine has been developed, which enables the quick and efficient synthesis of diarylated pyrimidines. The choice of solvent proved critical to the success of this reaction sequence, with alcoholic solvent mixtures affording
D W Roberts et al.
Chemical research in toxicology, 24(7), 1003-1011 (2011-06-16)
There is a strong impetus to develop nonanimal based methods to predict skin sensitization potency. An approach based on physical organic chemistry, whereby chemicals are classified into reaction mechanistic domains and quantitative models or read-across methods are derived for each
Charles Q Huang et al.
Bioorganic & medicinal chemistry letters, 14(9), 2083-2086 (2004-04-15)
A series of 2-dialkylamino-4-phenylpyrimidines (7) was designed and synthesized as CRF(1) antagonists. SAR studies of this series resulted in the discovery of potent and selective antagonists 7b and 7n bearing a 4-(2,4,6-trisubstituted-phenyl) ring and a bulky 2-(N-bis(cyclopropane)methyl-N-propyl)amino group.
Mahbub Alam et al.
Bioorganic & medicinal chemistry letters, 17(12), 3463-3467 (2007-04-27)
The development of a series of novel aminopyrimidines as inhibitors of c-Jun N-terminal kinases is described. The synthesis, in vitro inhibitory values for JNK1, JNK2 and CDK2, and the in vitro inhibitory value for a c-Jun cellular assay are discussed.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service