Assay
99%
refractive index
n20/D 1.5890 (lit.)
bp
314-315 °C (lit.)
density
1.115 g/mL
SMILES string
N#Cc1ccccc1N2CCNCC2
InChI
1S/C11H13N3/c12-9-10-3-1-2-4-11(10)14-7-5-13-6-8-14/h1-4,13H,5-8H2
InChI key
FRICBZWJFIRJOB-UHFFFAOYSA-N
Related Categories
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 3
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
118.0 °F - closed cup
Flash Point(C)
47.8 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service