Skip to Content
Merck
All Photos(1)

Documents

444804

Sigma-Aldrich

1-(2-Fluorophenyl)piperazine

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H13FN2
CAS Number:
Molecular Weight:
180.22
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

refractive index

n20/D 1.556 (lit.)

bp

150 °C/3 mmHg (lit.)

density

1.141 g/mL at 25 °C (lit.)

SMILES string

Fc1ccccc1N2CCNCC2

InChI

1S/C10H13FN2/c11-9-3-1-2-4-10(9)13-7-5-12-6-8-13/h1-4,12H,5-8H2

InChI key

IVTZRJKKXSKXKO-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

3, 4, 4-Trichloro-1-[4-(2-fluorophenyl) piperazinyl]-1-(n-hexadecylsulfanyl)-2-nitrobuta-1, 3-diene.
Ibis C, et al.
Acta Crystallographica Section E, Structure Reports Online, 62(3), o1147-o1148 (2006)
Collection of Czechoslovak Chemical Communications, 61, 1085-1085 (1996)
The Synthesis of Novel S-, S, S-, S, S, S-, and N, S-Substituted Nitrodienes from Polyhalonitrodienes.
Ibis C, et al.
Phosphorus, Sulfur, and Silicon and the Related Elements, 186(11), 2236-2249 (2011)
3, 4, 4-Trichloro-1-(4-chlorophenylsulfanyl)-1-[4-(2-fluorophenyl) piperazinyl]-2-nitrobuta-1, 3-diene.
4 Ibis C, et al.
Acta Crystallographica Section E, Structure Reports Online, 62(7), 2932-2933 (2006)
Canlan Jiang et al.
Water research, 106, 507-517 (2016-10-23)
Widespread occurrence of fluoroquinolone antibiotics (FQs) in surface water, groundwater, soil and sediment has been reported and their remediation is essentially needed. Sulfate radical (SO

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service