Skip to Content
Merck
All Photos(3)

Key Documents

562599

Sigma-Aldrich

Cesium chloride

99.99% trace metals basis

Synonym(s):

Cesium monochloride

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Linear Formula:
CsCl
CAS Number:
Molecular Weight:
168.36
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Assay:
99.99% trace metals basis
form:
powder

Quality Level

Assay

99.99% trace metals basis

form

powder

impurities

≤150.0 ppm Trace Metal Analysis

mp

645 °C (lit.)

SMILES string

[Cl-].[Cs+]

InChI

1S/ClH.Cs/h1H;/q;+1/p-1

InChI key

AIYUHDOJVYHVIT-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Cesium chloride is a white crystalline solid that is soluble in water. It is widely used in the field of solar cells and nucleic acid separation. It is partially soluble in ethanol and methanol but highly soluble in formic acid. Cesium chloride has a wide band gap of about 8.35 eV at 80 K.

Application

Cesium chloride can be used:
  • As a precursor to fabricate homogeneous perovskite films with high crystallinity and preferential orientation. These films enhance the power conversion efficiency of solar cells.[1]
  • To synthesize blue emission lead-free antimony Cesium halide Qdots of high PLQY of 46%[2] and Mn-Doped Cs2Na1–xAgxBiCl6 double perovskites for engineering the band gap and tailoring the energy transfer.[3]
  • To fabricate polyoxometalate electrode materials for Li-ion batteries. The addition of CsCl enhances the thermal stability and cycling stability of the battery cell.
  • To prepare a density gradient solution for the separation of nucleic acids.
Our Cesium chloride is highly prized for its exceptional purity of 99.99% in terms of trace metals. It is the preferred choice for synthesizing a wide range of perovskite materials.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

High Quantum Yield Blue Emission from Lead-Free Inorganic Antimony Halide Perovskite Colloidal Quantum Dots
Jian Zhang, et al.
ACS Nano, 11, 9294-9302 (2017)
Improvements in efficiency and stability of perovskite solar cells using a cesium chloride additive
Xiaodan Tang, et al.
ACS Applied Materials & Interfaces, 14, 26866-26872 (2022)
Homo- and Heterovalent Doping-Mediated Self-Trapped Exciton Emission and Energy Transfer in Mn-Doped Cs2Na1?xAgxBiCl6 Double Perovskites
Bao Ke, et al.
The Journal of Physical Chemistry Letters, 11, 340-348 (2020)
Tuan A Ho et al.
Langmuir : the ACS journal of surfaces and colloids, 28(2), 1256-1266 (2011-12-14)
All-atom molecular dynamics simulations were conducted to study the dynamics of aqueous electrolyte solutions confined in slit-shaped silica nanopores of various degrees of protonation. Five degrees of protonation were prepared by randomly removing surface hydrogen atoms from fully protonated crystalline
Piotr Jurkiewicz et al.
Biochimica et biophysica acta, 1818(3), 609-616 (2011-12-14)
Effects of alkali metal chlorides on the properties of mixed negatively charged lipid bilayers are experimentally measured and numerically simulated. Addition of 20mol% of negatively charged phosphatidylserine to zwitterionic phosphatidylcholine strengthens adsorption of monovalent cations revealing their specificity, in the

Articles

Colloidal quantum dots (CQDs) are semiconducting crystals of only a few nanometers (ca. 2–12 nm) coated with ligand/surfactant molecules to help prevent agglomeration.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service