Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Rubrene
Synonym(s):
5,6,11,12-Tetraphenylnaphthacene
Empirical Formula (Hill Notation):
C
42
H
28
CAS Number:
517-51-1
Molecular Weight:
532.67
EC Number:
208-242-0
Products
Brand
Sigma-Aldrich (4)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Special Grade
Sublimed grade (1)
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
554073
Rubrene,
≥98%
Expand
View Pricing
551112
Rubrene,
sublimed grade, 99.99% trace metals basis
Expand
View Pricing
932450
Rubrene,
≥99% (HPLC)
Expand
View Pricing
R2206
Rubrene,
powder
Expand
View Pricing