Skip to Content
Merck
All Photos(1)

Key Documents

548618

Sigma-Aldrich

2-Benzyl-4-chlorophenol

95%

Synonym(s):

Chlorophene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2C6H3(Cl)OH
CAS Number:
Molecular Weight:
218.68
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

bp

160-162 °C (lit.)

mp

46-49 °C (lit.)

density

1.188 g/mL at 25 °C (lit.)

functional group

chloro
phenyl

SMILES string

Oc1ccc(Cl)cc1Cc2ccccc2

InChI

1S/C13H11ClO/c14-12-6-7-13(15)11(9-12)8-10-4-2-1-3-5-10/h1-7,9,15H,8H2

InChI key

NCKMMSIFQUPKCK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Dam. 1 - Repr. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 2 Oral

Target Organs

Kidney

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

No data available

Flash Point(C)

No data available

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Allergic contact dermatitis from orthobenzyl parachlorophenol in a drinking glass cleaner.
T S Sonnex et al.
Contact dermatitis, 14(4), 247-248 (1986-04-01)
L R Kao et al.
Journal of toxicology and environmental health, 18(3), 441-458 (1986-01-01)
The disposition and metabolism of o-benzyl-p-chlorophenol (BCP) were studied in male Fischer-344 rats. Three days after oral administration of [14C]BCP at 10, 100, or 1000 mg/kg, more than 90% of each dose was excreted in urine and feces. Comparison of
Gabriela Elizabeth Quintanilla-Villanueva et al.
Biosensors, 11(2) (2021-02-13)
Chlorophene is an important antimicrobial agent present in disinfectant products which has been related to health and environmental effects, and its detection has been limited to chromatographic techniques. Thus, there is a lack of research that attempts to develop new
Beatriz Merchel Piovesan Pereira et al.
Frontiers in microbiology, 12, 640923-640923 (2021-03-16)
Biocide use is essential and ubiquitous, exposing microbes to sub-inhibitory concentrations of antiseptics, disinfectants, and preservatives. This can lead to the emergence of biocide resistance, and more importantly, potential cross-resistance to antibiotics, although the degree, frequency, and mechanisms that give
L R Kao et al.
Biochemical pharmacology, 35(4), 613-620 (1986-02-15)
o-Benzyl-p-chlorophenol (BCP) is widely used as a broad spectrum disinfectant. Treatment of male Fischer 344 rats with BCP resulted in an increase in cytochrome P-450 content and an accompanying decrease in aryl hydrocarbon hydroxylase (AHH) activity in both liver and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service