Skip to Content
Merck
All Photos(1)

Key Documents

540080

Sigma-Aldrich

(S)-(+)-Epichlorohydrin

98%

Synonym(s):

(S)-(+)-2-(Chloromethyl)oxirane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H5ClO
CAS Number:
Molecular Weight:
92.52
Beilstein:
1420784
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

vapor density

3.2 (vs air)

vapor pressure

12.5 mmHg ( 20 °C)

Assay

98%

optical activity

[α]20/D +35°, c = 1 in methanol

optical purity

ee: 97% (GLC)

autoignition temp.

1421 °F

expl. lim.

21 %

refractive index

n20/D 1.438 (lit.)

bp

92-93 °C/360 mmHg (lit.)

density

1.183 g/mL at 25 °C (lit.)

SMILES string

ClC[C@@H]1CO1

InChI

1S/C3H5ClO/c4-1-3-2-5-3/h3H,1-2H2/t3-/m1/s1

InChI key

BRLQWZUYTZBJKN-GSVOUGTGSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Chiral building block in the enantioselective syntheses of hydroxyisoxazolidines and (+)-cis-sylvaticin, a potential antitumor agent.
Used in the total synthesis of Macquarimicins and in the total synthesis of the macrolide RK-397.
Used to synthesize inhibitors of fatty acid oxidation as potential metabolic modulators.

Legal Information

Manufactured under license by Sterling Pharma Solutions Limited, using Jacobsen HKR technology.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Carc. 1B - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

89.6 °F - closed cup

Flash Point(C)

32 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron Letters, 47, 7635-7635 (2006)
Ryosuke Munakata et al.
Journal of the American Chemical Society, 125(48), 14722-14723 (2003-12-04)
The first total synthesis of (+)-macquarimicin A (1), a novel inhibitor of neutral sphingomyelinase (N-SMase) with antiinflammatory activity, has been accomplished. The present work determined the absolute configuration of (+)-1 and revised the C(2)-C(3) geometry to be Z. The synthesis
Elfatih Elzein et al.
Bioorganic & medicinal chemistry letters, 14(4), 973-977 (2004-03-12)
We describe the synthesis of novel inhibitors of fatty acid oxidation as potential metabolic modulators for the treatment of stable angina. Replacement of the 2H-benzo[d]1,3-dioxolene ring system in our initial lead 3 with different benzthiazoles, benzoxazoles and introducing small alkyl
Svetlana A Burova et al.
Journal of the American Chemical Society, 126(8), 2495-2500 (2004-02-26)
The total synthesis of the natural product RK-397 is based on a new synthetic strategy for assembling polyacetate structures, by efficient cross-coupling of nucleophilic terminal alkyne modules with electrophilic epoxides bearing another alkyne at the opposite terminus. The natural product
Ryosuke Munakata et al.
Journal of the American Chemical Society, 126(36), 11254-11267 (2004-09-10)
A total synthesis of the macquarimicins A-C (1-3), novel natural products with intriguing tetra- or pentacyclic frameworks, has been achieved. The synthesis features an extensive investigation of the biosynthesis-based intramolecular Diels-Alder (IMDA) reactions of (E,Z,E)-1,6,8-nonatrienes. Considering possible biosynthetic sequences, four

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service