Skip to Content
Merck
All Photos(1)

Key Documents

E1012

Sigma-Aldrich

Epibromohydrin

98%

Synonym(s):

1-Bromo-2,3-epoxypropane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H5BrO
CAS Number:
Molecular Weight:
136.98
Beilstein:
79786
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.482 (lit.)

bp

134-136 °C (lit.)

mp

−40 °C (lit.)

density

1.601 g/mL at 25 °C (lit.)

SMILES string

BrCC1CO1

InChI

1S/C3H5BrO/c4-1-3-2-5-3/h3H,1-2H2

InChI key

GKIPXFAANLTWBM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Epibromohydrin can be used:
  • As an alkylating agent.
  • As a reactant in the multistep synthesis of analogs of febrifugine and P7C3.
  • To synthesize cyclic carbonates by coupling with CO2 in the presence of bis(triphenylphosphine)immium(PPN)-manganese carbonyl catalyst.
  • For the synthesis of triazole-oxazolidinones possessing antibacterial activities.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Carc. 2 - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

138.2 °F - closed cup

Flash Point(C)

59 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cyclin-dependent kinase 4 inhibitors as a treatment for cancer. Part 1: identification and optimisation of substituted 4, 6-bis anilino pyrimidines.
Beattie JF, et al.
Bioorganic & Medicinal Chemistry, 13(18), 2955-2960 (2003)
Development of proneurogenic, neuroprotective small molecules.
MacMillan KS, et al.
Journal of the American Chemical Society, 133(5), 1428-1437 (2011)
Coupling reactions of CO2 with neat epoxides catalyzed by PPN salts to yield cyclic carbonates.
Sit WN, et al.
The Journal of Organic Chemistry, 70(21), 8583-8586 (2005)
Synthesis of triazole-oxazolidinones via a one-pot reaction and evaluation of their antimicrobial activity.
Demaray JA, et al.
Bioorganic & Medicinal Chemistry Letters, 18(17), 4868-4871 (2008)
Imidazo [1, 2-a] pyridines: a potent and selective class of cyclin-dependent kinase inhibitors identified through structure-based hybridisation.
Anderson M, et al.
Bioorganic & Medicinal Chemistry, 13(18), 3021-3026 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service