Skip to Content
Merck
All Photos(1)

Documents

539899

Sigma-Aldrich

Bis(pentafluorophenyl) carbonate

97%, for peptide synthesis

Synonym(s):

Pentafluorophenyl carbonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6F5O)2CO
CAS Number:
Molecular Weight:
394.12
Beilstein:
2029727
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

product name

Bis(pentafluorophenyl) carbonate, 97%

Assay

97%

reaction suitability

reaction type: Carbonylations

mp

47-50 °C (lit.)

application(s)

peptide synthesis

SMILES string

Fc1c(F)c(F)c(OC(=O)Oc2c(F)c(F)c(F)c(F)c2F)c(F)c1F

InChI

1S/C13F10O3/c14-1-3(16)7(20)11(8(21)4(1)17)25-13(24)26-12-9(22)5(18)2(15)6(19)10(12)23

InChI key

IOVVFSGCNWQFQT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Bis(pentafluorophenyl) carbonate is the equivalent of carbonyl compound generally used in coupling reactions. It is used as a reagent in the preparation of azapeptides.

Application

Bis(pentafluorophenyl) carbonate can be used in the preparation of:
  • A cyclic carbonate named 6,6′-(ethane-1,2-diyl)bis(1,3,6-dioxazocan-2-one), which is a key intermediate for the synthesis of non-isocyanate polyurethanes.
  • A coumarin based aliphatic polycarbonate named 5-(4-methylumbelliferyloxycarbonyl)-5-methyl-1,3-dioxan-2-one (MUC).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Bis(pentafluorophenyl)carbonate
Encyclopedia of Reagents for Organic Synthesis, Second Edition (2004)
V N Medvedkin et al.
Bioorganicheskaia khimiia, 15(4), 460-464 (1989-04-01)
Dipentafluorophenylcarbonate, belonging to transesterifiying reagents, has been prepared and used for the synthesis of pentafluorophenyl esters of amino acids. In contrast to many other reagents of the kind, its preparation is simple, it is highly reactive and at the same
Room temperature synthesis of non-isocyanate polyurethanes (NIPUs) using highly reactive N-substituted 8-membered cyclic carbonates
Yuen A, et al.
Polym. Chem., 7(11), 2105-2111 (2016)
Synthesis of cinnamoyl and coumarin functionalized aliphatic polycarbonates
Chesterman JP, et al.
Polym. Chem., 8(48), 7515-7528 (2017)
[Dipentafluorophenylcarbonate in the synthesis of oligonucleotides by the H-phosphonate method].
V A Efimov et al.
Bioorganicheskaia khimiia, 20(3), 323-326 (1994-03-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service