Skip to Content
Merck
All Photos(3)

Key Documents

377074

Sigma-Aldrich

Pentafluorophenyl trifluoroacetate

98%, for peptide synthesis

Synonym(s):

Trifluoroacetic acid pentafluorophenyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3CO2C6F5
CAS Number:
Molecular Weight:
280.07
Beilstein:
2003848
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

Product Name

Pentafluorophenyl trifluoroacetate, 98%

Assay

98%

form

liquid

refractive index

n20/D 1.368 (lit.)

bp

122-123 °C (lit.)

density

1.63 g/mL at 25 °C (lit.)

application(s)

peptide synthesis

functional group

ester
fluoro

SMILES string

Fc1c(F)c(F)c(OC(=O)C(F)(F)F)c(F)c1F

InChI

1S/C8F8O2/c9-1-2(10)4(12)6(5(13)3(1)11)18-7(17)8(14,15)16

InChI key

VCQURUZYYSOUHP-UHFFFAOYSA-N

Application

Pentafluorophenyl trifluoroacetate can be used:
  • In the esterification of 2′-carboxyrhodamine dye to form pentafluorophenyl ester as a single isomer.
  • As an acylating agent and a coupling agent for the peptide-type coupling of N-substituted glycine oligomers.
  • In the synthesis of pentafluorophenyl thiophene-3-acetate (PFPTA) by reacting with 3-thiopheneacetic acid.

Other Notes

may contain a small amount of pentafluorophenol

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

125.6 °F - closed cup

Flash Point(C)

52 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Unexpected reactivity of the 2′-carboxyl functionality in rhodamine dyes.
Haack RA, et al.
Tetrahedron Letters, 58(18), 1733-1737 (2017)
Vancomycin-conjugated polythiophene for the detection and imaging of Gram-positive bacteria.
Ning LG, et al.
Journal of Material Chemistry B: Materials for Biology and Medicine, 5(44), 8814-8820 (2017)
Nathan J Van Zee et al.
Nature, 558(7708), 100-103 (2018-06-01)
Water directs the self-assembly of both natural1,2 and synthetic3-9 molecules to form precise yet dynamic structures. Nevertheless, our molecular understanding of the role of water in such systems is incomplete, which represents a fundamental constraint in the development of supramolecular
Weak backbone CH?O=C and side chain t Bu?t Bu London interactions help promote helix folding of achiral NtBu peptoids.
Angelici G, et al.
Chemical Communications (Cambridge, England), 52(24), 4573-4576 (2016)
Rory K Morgan et al.
ACS chemical biology, 10(8), 1778-1784 (2015-05-16)
ADP-ribosylation is essential for cell function, yet there is a dearth of methods for detecting this post-translational modification in cells. Here, we describe a clickable aminooxy alkyne (AO-alkyne) probe that can detect cellular ADP-ribosylation on acidic amino acids following Cu-catalyzed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service