Skip to Content
Merck
All Photos(1)

Key Documents

534803

Sigma-Aldrich

Amano Lipase M from Mucor javanicus

≥10,000 U/g, pH 7.0, 40 °C (Optimum pH and temperature)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

specific activity

≥10,000 U/g, pH 7.0, 40 °C (Optimum pH and temperature)

storage temp.

2-8°C

InChI

1S/C11H9N3O2.Na/c15-8-4-5-9(10(16)7-8)13-14-11-3-1-2-6-12-11;/h1-7,16H,(H,12,14);/q;+1/b13-9-;

InChI key

QWZUIMCIEOCSJF-CHHCPSLASA-N

Application

Amano Lipase M from Mucor javanicus has been used:
  • as a component in calcium alginate beads or immobilization to study its efficiency and stability in the biocatalytic synthesis of butyl butyrate in aqueous media[1]
  • as a component in calcium alginate beads for transmission electron microscope (TEM) imaging[1]
  • in immobilization to study its efficiency in ester synthesis and to evaluate its catalytic behavior[2]
  • in catalyzing Michael addition reactions of pyrimidine derivatives to disaccharide acrylates[3]

Biochem/physiol Actions

Amano lipase M is capable of catalyzing Michael addition of pyrimidine with disaccharide acrylates in organic media.[3]

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Biolubricant production under zero-waste Moringa oleifera Lam biorefinery approach for boosting circular economy
Barbosa MS, et al.
Industrial Crops and Products, 113542-113542 (2021)
Hong-Xia Dai et al.
Biotechnology letters, 28(18), 1503-1507 (2006-09-07)
The Michael addition of fluorouracil (0.01 mol), uracil (0.01 mol), thymine (0.01 mol) to disaccharide acrylates (0.03 mol) was carried out in pyridine (20 ml) at 50 degrees C for 72 h with lipase M from Mucor javanicus (200 mg).
Mika Henrikki Sipponen et al.
Nature communications, 9(1), 2300-2300 (2018-06-14)
Dehydration reactions proceed readily in water-filled biological cells. Development of biocatalysts that mimic such compartmentalized reactions has been cumbersome due to the lack of low-cost nanomaterials and associated technologies. Here we show that cationic lignin nanospheres function as activating anchors

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service