Skip to Content
Merck
All Photos(1)

Key Documents

532851

Sigma-Aldrich

Methyl 2-iodobenzoate

97%

Synonym(s):

Methyl o-iodobenzoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
IC6H4CO2CH3
CAS Number:
Molecular Weight:
262.04
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.604 (lit.)

bp

149-150 °C/10 mmHg (lit.)

density

1.784 g/mL at 25 °C (lit.)

SMILES string

COC(=O)c1ccccc1I

InChI

1S/C8H7IO2/c1-11-8(10)6-4-2-3-5-7(6)9/h2-5H,1H3

InChI key

BXXLTVBTDZXPTN-UHFFFAOYSA-N

General description

Methyl 2-iodobenzoate can be prepared from 2-iodobenzoic acid via esterification. It undergoes cobalt-catalyzed cyclization with aldehydes to form phthalide derivatives. The microbial dihydroxylation of methyl 2-iodobenzoate forms a nonracemic iodocyclohexene carboxylate intermediate. This intermediate forms the precursor for preparing kibdelone C.

Application

Methyl 2-iodobenzoate may be used in the preparation of:
  • N-substituted 4-methylene-3,4-dihydro-1(2H)-isoquinolin-1-ones
  • methyl diphenylacetylene-2-carboxylate
  • methyl 2-heptynylphenylbenzoate
  • (E)-2-[3-[3-[2-(7-chloro-2-quinolinyl)ethenyl]phenyl]-3-oxopropyl]benzoic acid methyl ester
  • 4-(2-carbomethoxyphenyl)-3-(1-methylethoxy)cyclobut-3-ene-1,2-dione
  • 3-(2-carbomethoxyphenyl)-4-methylcyclobuten-3-ene-1,2-dione 2-(ethylene acetal)

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An efficient synthesis of LTD4 antagonist L-699,392.
The Journal of Organic Chemistry, 58(14), 3731-3735 (1993)
Highly Efficient Cyclization of o?Iodobenzoates with Aldehydes Catalyzed by Cobalt Bidentate Phosphine Complexes: A Novel Entry to Chiral Phthalides.
Chang HT, et al.
Chemistry (Weinheim An Der Bergstrasse, Germany), 13(15), 4356-4363 (2007)
3-Stannylcyclobutenediones as nucleophilic cyclobutenedione equivalents. Synthesis of substituted cyclobutenediones and cyclobutenedione monoacetals and the beneficial effect of catalytic copper iodide on the Stille reaction.
Liebeskind LS and Fengl RW.
The Journal of Organic Chemistry, 55(19), 5359-5364 (1990)
Palladium homogeneous and supported catalysis: synthesis of functional acetylenics and cyclisation to heterocycles.
Villemin D and Goussu D.
Heterocycles, 29(7), 1255-1261 (1989)
Synthesis of N-substituted 4-methylene-3, 4-dihydro-1 (2H)-isoquinolin-1-ones via a palladium-catalysed three-component process.
Grigg R, et al.
Tetrahedron, 43(14), 2601-2603 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service