Skip to Content
Merck
All Photos(3)

Key Documents

529575

Sigma-Aldrich

Cadmium chloride hydrate

99.995% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CdCl2.xH2O
CAS Number:
Molecular Weight:
183.32 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

vapor density

6.3 (vs air)

Quality Level

Assay

99.995% trace metals basis

form

solid

reaction suitability

reagent type: catalyst
core: cadmium

impurities

≤55.0 ppm Trace Metal Analysis

density

3.327 g/mL at 25 °C (lit.)

SMILES string

O.Cl[Cd]Cl

InChI

1S/Cd.2ClH.H2O/h;2*1H;1H2/q+2;;;/p-2

InChI key

OISMQLUZKQIKII-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Muta. 1B - Repr. 1B - STOT RE 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Leena Sivula et al.
Chemosphere, 210, 304-311 (2018-07-14)
Increasing metal consumption is driving the introduction of new techniques such as biomining to exploit low grade ores. The biomining impacts notably aquatic ecosystems, yet, the applicability of ecotoxicological tests to study the complex mixture effects of mining waters is
Olga Guselnikova et al.
Sensors (Basel, Switzerland), 19(9) (2019-05-10)
In this contribution, we propose a novel functional surface-enhanced Raman spectroscopy (SERS) platform for the detection of one of the most hazardous heavy metal ions, Hg2+. The design of the proposed sensor is based on the combination of surface plasmon-polariton
Alan Kania et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 40(28), 5362-5375 (2020-06-14)
Binge-eating disorder is the most common eating disorder. Various neuropeptides play important roles in the regulation of feeding behavior, including relaxin-3 (RLN3), which stimulates food intake in rats through the activation of the relaxin-family peptide-3 receptor (RXFP3). Here we demonstrate
Beth A McNally et al.
The Journal of physiology, 598(9), 1775-1790 (2019-06-10)
Circadian oscillations in spontaneous action potential firing in the suprachiasmatic nucleus (SCN) translate time-of-day throughout the mammalian brain. The ion channels that regulate the circadian pattern of SCN firing have not been comprehensively identified. Ca2+ channels regulate action potential activity
Meritxell Navarro-Reig et al.
Analytical and bioanalytical chemistry, 407(29), 8835-8847 (2015-09-26)
The comprehensive analysis of untargeted metabolomics data acquired using LC-MS is still a major challenge. Different data analysis tools have been developed in recent years such as XCMS (various forms (X) of chromatography mass spectrometry) and multivariate curve resolution alternating

Articles

The diversity of applications and nanostructured materials accessible using ultrasonic spray methods are highlighted in this article.

The diversity of applications and nanostructured materials accessible using ultrasonic spray methods are highlighted in this article.

The diversity of applications and nanostructured materials accessible using ultrasonic spray methods are highlighted in this article.

The diversity of applications and nanostructured materials accessible using ultrasonic spray methods are highlighted in this article.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service