Skip to Content
Merck
All Photos(4)

Key Documents

268690

Sigma-Aldrich

Lead(II) chloride

powder, 98%

Synonym(s):

Cotunnite

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
PbCl2
CAS Number:
Molecular Weight:
278.11
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
powder

grade

for analytical purposes

vapor pressure

1 mmHg ( 547 °C)

Assay

98%

form

powder

reaction suitability

reagent type: catalyst
core: lead

bp

950 °C (lit.)

mp

501 °C (lit.)

density

5.85 g/mL at 25 °C (lit.)

SMILES string

Cl[PbH2]Cl

InChI

1S/2ClH.Pb/h2*1H;/q;;+2/p-2

InChI key

HWSZZLVAJGOAAY-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Lead(II) chloride (PbCl2) (98%) is a lead halide that can be used in the formation of a precursor solution to form perovskites films. These films and sheets can be used in the fabrication of hybrid solar absorbers and photodetectors. It also has potential application as a thermoelectric material.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Repr. 1A - STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Methylammonium lead mixed halide films processed with a new composition for planar perovskite solar cells.
Park BS, et al.
Applied Surface Science, 427, 421-426 (2018)
Strong Quantum Confinement and Fast Photoemission Activation in CH3NH3PbI3 Perovskite Nanocrystals Grown within Periodically Mesostructured Films.
Anaya M, et al.
Advanced Optical Materials, 5(8) (2017)
Md Kawser Ahmed et al.
Environmental monitoring and assessment, 182(1-4), 197-204 (2011-01-07)
The present investigation was undertaken to study the induction of DNA damage by lead chloride (PbCl(2)) in freshwater climbing perch Anabas testudineus using alkaline single cell gel electrophoresis (comet assay). Based on the LC(50) values of lead chloride of A.
Travis Rush et al.
Neurotoxicology, 30(1), 47-51 (2008-11-26)
Chelation therapy for the treatment of acute, high dose exposure to heavy metals is accepted medical practice. However, a much wider use of metal chelators is by alternative health practitioners for so called "chelation therapy". Given this widespread and largely
Issa Tapsoba et al.
Analytical chemistry, 82(2), 457-460 (2009-12-25)
Lead-based compounds were used during antiquity as both pigments and medicines in the formulation of makeup materials. Chemical analysis of cosmetics samples found in Egyptians tombs and the reconstitution of ancient recipes as reported by Greco-Roman authors have shown that

Articles

Colloidal quantum dots (CQDs) are semiconducting crystals of only a few nanometers (ca. 2–12 nm) coated with ligand/surfactant molecules to help prevent agglomeration.

Controlled synthesis of metal clusters regulates ligands and atoms, advancing metal nanomaterial synthesis.

Controlled synthesis of metal clusters regulates ligands and atoms, advancing metal nanomaterial synthesis.

Controlled synthesis of metal clusters regulates ligands and atoms, advancing metal nanomaterial synthesis.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service