Skip to Content
Merck
All Photos(1)

Key Documents

527483

Sigma-Aldrich

Poly(sodium 4-styrenesulfonate) solution

average Mw ~70,000, 30 wt. % in H2O

Synonym(s):

PSS, Poly(4-styrenesulfonic acid) sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C8H7NaO3S)n
CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

mol wt

average Mw ~70,000

Quality Level

concentration

30 wt. % in H2O

refractive index

n20/D 1.395

bp

100 °C

density

1.158 g/mL at 25 °C

SMILES string

[S](=O)(=O)(O)c1ccc(cc1)C=C

InChI

1S/C8H8O3S/c1-2-7-3-5-8(6-4-7)12(9,10)11/h2-6H,1H2,(H,9,10,11)

InChI key

MAGFQRLKWCCTQJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Effective antistatic and specialty dispersant.

Features and Benefits

Anionic polyelectrolyte

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Johannes Frueh et al.
The journal of physical chemistry. B, 116(40), 12257-12262 (2012-09-18)
Polyelectrolyte (PE) multilayer (PEM) thin films prepared by layer-by-layer self-assembly on flexible substrates are exposed to elongation in many fields of technology. Upon elongation, these types of films are showing interesting, but not understood, phenomena, such as controlled wetting, stimuli-responsive
Ahmed Chaaban et al.
Renal failure, 35(2), 185-188 (2013-02-14)
There is insufficient evidence on the utility of potassium-binding resins in patients with end-stage renal disease on dialysis. In addition, their poor tolerability raises concerns of patient adherence. We aimed to assess the efficacy of calcium resonium and investigate the
Jean-Christy F Cameron et al.
Paediatric drugs, 15(1), 43-48 (2013-01-19)
In pediatric patients at risk of hyperkalemia there are limited treatment or preventive alternatives for this electrolyte imbalance. Oral or rectal sodium polystyrene sulfonate (SPS) has several potential adverse effects, and dietary potassium restriction may compromise nutrition. Pretreatment of infant
Johannes Frueh et al.
Physical chemistry chemical physics : PCCP, 15(2), 483-488 (2012-11-23)
Polyelectrolyte multilayers (PEMs) deposited on flexible supports are promising candidates for many applications ranging from controlled wettability over stimuli responsive nanovalves to lithography free surface structuring. Since many potential applications involve elongation of these films, we investigated the effect of
S Oughlis et al.
Journal of biomedical materials research. Part A, 101(2), 582-589 (2012-09-11)
Osseointegration of metallic implants used in orthopedic surgery requires that osteoprogenitor cells attach and adhere to the surface, then proliferate, differentiate into osteoblasts, and finally produce mineralized matrix. Because the ability of progenitor cells to attach to a scaffold surface

Articles

We present an article that discusses two applications in particular; first, using these layers as polyelectrolyte membranes to control permeability.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service