Skip to Content
Merck
All Photos(1)

Key Documents

526274

Sigma-Aldrich

5-Fluorosalicylaldehyde

97%

Synonym(s):

5-Fluoro-2-hydroxybenzaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H3(OH)CHO
CAS Number:
Molecular Weight:
140.11
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

82-85 °C (lit.)

functional group

aldehyde
fluoro

SMILES string

Oc1ccc(F)cc1C=O

InChI

1S/C7H5FO2/c8-6-1-2-7(10)5(3-6)4-9/h1-4,10H

InChI key

FDUBQNUDZOGOFE-UHFFFAOYSA-N

General description

5-Fluorosalicylaldehyde can be synthesized from 4-fluorophenol.

Application

5-Fluorosalicylaldehyde may be used to synthesize:
  • managanese (III) complex [MnLa1,3-N3)]n [H2La = N,N′-bis(5-fluorosalicylidene)-1,2-diaminoethane]
  • enantiopure 4-amino-6-fluoro-3-(hydroxymethyl)chromanes
  • 4,4′-difluoro-2,2′-[(hydrazine-1,2-diylidene)bis(methanylylidene)]-diphenol
  • N,N′-bis(5-fluoro-2-hydroxybenzylidene)ethylenediamine
  • N,N′-(2-hydroxypropane-1,3-diyl)-bis(salicylaldimine (H2L)

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Large perturbations of long-range n J (1H, 1H) and n J (1H, 19F) by the intramolecular hydrogen bonds in 2-mercaptobenzaldehyde, salicylaldehyde, and some derivatives. Reference structures for intramolecular hydrogen bonds.
Schaefer T, et al.
Canadian Journal of Chemistry, 71(7), 960-967 (1993)
Synthesis, Characterization and Crystal Structures of N, N'-bis (5-Fluoro-2-hydroxybenzylidene)-ethylenediamine and 4, 4'-Difluoro-2, 2'-[(hydrazine-1, 2-diylidene) bis (methanylylidene)] diphenol.
Zhang WM, et al.
Asian Journal of Chemistry, 26(22), 7843-7843 (2014)
Manganese (III) complexes derived from bis-Schiff bases: Synthesis, structures, and antimicrobial activity.
Liu QR, et al.
Russian Journal of Coordination Chemistry, 40(10), 757-763 (2014)
Synthesis and crystal structure of a novel tri-nuclear nickel (II) complex derived from N, N'-(2-hydroxypropane-1, 3-diyl) bis (salicylaldimine).
Cui YM, et al.
Russian Journal of Coordination Chemistry, 40(10), 768-772 (2014)
Synthesis of Enantiopure 4-Amino-6-fluoro-3-(hydroxymethyl) chromanes via Intramolecular Nitrone Cycloadditions.
Broggini G, et al.
J. Chem. Res. Synop., 2, 36-37 (1997)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service