510718
Titanium(IV) butoxide, polymer
Synonym(s):
Tetrabutyl titanate polymer, Tilcom PBT, Titanium tetrabutoxide polymer, Tytan PBT, Tyzor BTP, Tyzor PB, Vertec PBT
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
CAS Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
vapor pressure
<0.8 mmHg ( 20 °C)
Quality Level
reaction suitability
core: titanium
reagent type: catalyst
mp
−39 °C (lit.)
density
1.13 g/mL at 25 °C (lit.)
SMILES string
CCCCO[Ti](OCCCC)(OCCCC)OCCCC
InChI
1S/4C4H9O.Ti/c4*1-2-3-4-5;/h4*2-4H2,1H3;/q4*-1;+4
InChI key
YHWCPXVTRSHPNY-UHFFFAOYSA-N
Related Categories
Features and Benefits
Used in the study of thermal properties of siloxane coating KT-30 produced in varying methods.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Flam. Liq. 3
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
89.6 °F - closed cup
Flash Point(C)
32 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Hye Yun Jeong et al.
ACS nano, 10(1), 770-777 (2015-12-10)
While transmission electron microscopy and scanning tunneling microscopy reveal atomic structures of point defect and grain boundary in monolayer transition-metal dichalcogenides (TMDs), information on point defect distribution in macroscale is still not available. Herein, we visualize the point defect distribution
S D Makeev et al.
Klinicheskaia khirurgiia, (1)(1), 19-20 (1989-01-01)
By the method of tissue culture, it is established that the capron threads stained by brilliant green, coated with the chemosorbed substances on the base of titanium alkoxides, polyalkylhydridsiloxanes and polyorganilsiloxanes have no histotoxic properties and can be used in
Jun-Bing Fan et al.
Chemical communications (Cambridge, England), 46(31), 5805-5807 (2010-07-01)
A low cost and environmentally benign process for tetrabutyl titanate-controlled ring-opening polymerization of epsilon-caprolactone at ambient temperature of 10-40 degrees C is introduced affording poly(epsilon-caprolactone) with pre-designed molecular weights and narrow molecular weight distribution.
Beidou Xi et al.
Huan jing ke xue= Huanjing kexue, 23(1), 66-69 (2002-05-04)
Photocatalysts Pt/TiO2 coated on hollow glass beads were prepared by tetrabutyl titanate hydrolysis with Sodium silicate on hollow glass beads at various condition and loaded with platinum varying from 0.2% to 2.4% by weight. Sodium pentachlorophenolate (PCP-Na) solution were used
S Anantharaj et al.
Biomacromolecules, 13(8), 2446-2455 (2012-06-21)
A new dual ester-urethane melt condensation methodology for biological monomers-amino acids was developed to synthesize new classes of thermoplastic polymers under eco-friendly and solvent-free polymerization approach. Naturally abundant L-amino acids were converted into dual functional ester-urethane monomers by tailor-made synthetic
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service