Skip to Content
Merck
All Photos(1)

Key Documents

50280

Sigma-Aldrich

Gly-His hydrochloride hydrate

≥99.0% (calc. based on dry substance, AT)

Synonym(s):

Glycyl-L-histidine hydrochloride hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H12N4O3 · HCl · xH2O
CAS Number:
Molecular Weight:
248.67 (anhydrous basis)
Beilstein:
3743779
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥99.0% (calc. based on dry substance, AT)

optical activity

[α]20/D +31.5±1.0°, c = 2% in H2O (dry matter)

reaction suitability

reaction type: solution phase peptide synthesis

impurities

~1 mol water

application(s)

peptide synthesis

SMILES string

O.Cl.NCC(=O)N[C@@H](Cc1c[nH]cn1)C(O)=O

InChI

1S/C8H12N4O3.ClH.H2O/c9-2-7(13)12-6(8(14)15)1-5-3-10-4-11-5;;/h3-4,6H,1-2,9H2,(H,10,11)(H,12,13)(H,14,15);1H;1H2/t6-;;/m0../s1

InChI key

VGVLXWZHJBRECR-ILKKLZGPSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N Manoj et al.
The journal of peptide research : official journal of the American Peptide Society, 56(4), 210-217 (2000-11-18)
The crystal structures of the complexes of oxalic acid with glycyl-L-histidine and L-histidyl-L-alanine were determined. The three crystallographically independent peptide molecules in the complexes have closed conformations. The terminal carboxyl group of the dipeptide and the oxalate ion in the
Flora Carrera et al.
Inorganic chemistry, 43(21), 6674-6683 (2004-10-13)
Knowledge of the complexes formed by N-coordinating ligands and Cu(II) ions is of relevance in understanding the interactions of this ion with biomolecules. Within this framework, we investigated Cu(II) complexation with mono- and polydentate ligands, such as ammonia, ethylenediamine (en)
Girolamo Casella et al.
Inorganic chemistry, 47(11), 4796-4807 (2008-05-08)
We have tested several computational protocols, at the nonrelativistic DFT level of theory, for the calculation of 1J(119Sn, 13C) and 2J(119Sn, 1H) spin-spin coupling constants in di- and trimethyltin(IV) derivatives with various ligands. Quite a good agreement with experimental data
Michael S Kostelansky et al.
Biochemistry, 43(9), 2475-2483 (2004-03-03)
Structural analysis of recombinant fibrinogen fragment D revealed that the calcium-binding site (beta2-site) composed of residues BbetaAsp261, BbetaAsp398, BbetaGly263, and gammaGlu132 is modulated by the "B:b" interaction. To determine the beta2-site's role in polymerization, we engineered variant fibrinogen gammaE132A in
Giampiero De Sanctis et al.
Journal of biological inorganic chemistry : JBIC : a publication of the Society of Biological Inorganic Chemistry, 11(2), 153-167 (2005-12-13)
The pH dependence of redox properties, spectroscopic features and CO binding kinetics for the chelated protohemin-6(7)-L-histidine methyl ester (heme-H) and the chelated protohemin-6(7)-glycyl-L-histidine methyl ester (heme-GH) systems has been investigated between pH 2.0 and 12.0. The two heme systems appear

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service