Skip to Content
Merck
All Photos(1)

Key Documents

499862

Sigma-Aldrich

Tributyl(1-propynyl)tin

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)3]3SnC≡CCH3
CAS Number:
Molecular Weight:
329.11
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

refractive index

n20/D 1.483 (lit.)

bp

277 °C (lit.)

density

1.082 g/mL at 25 °C (lit.)

SMILES string

CCCC[Sn](CCCC)(CCCC)C#CC

InChI

1S/3C4H9.C3H3.Sn/c3*1-3-4-2;1-3-2;/h3*1,3-4H2,2H3;1H3;

InChI key

KCQJLTOSSVXOCC-UHFFFAOYSA-N

Application

Tributyl(1-propynyl)tin can be used:
  • As a starting material in the synthesis of phenyl-substituted pyrimidine-dione by reacting with ethyl isocyanate via cycloaddition and Stille coupling reactions.
  • In the synthesis of 18F labeled triazolo quinoline derivative, a positron emission tomography (PET) ligand used in the in vivo imaging of metabotropic glutamate receptor type 1.
  • In one of the key synthetic steps for the synthesis of 4-(1-aryltriazol-4-yl)-tetrahydropyridines as potent mGluR1 antagonists.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and evaluation of 6-[1-(2-[18F] fluoro-3-pyridyl)-5-methyl-1H-1,2,3-triazol-4-yl]quinoline for positron emission tomography imaging of the metabotropic glutamate receptor type 1 in brain
Fujinaga M, et al.
Bioorganic & Medicinal Chemistry, 19(1), 102-110 (2011)
Discovery and biological profile of 4-(1-aryltriazol-4-yl)-tetrahydropyridines as an orally active new class of metabotropic glutamate receptor 1 antagonist
Ito S, et al.
Bioorganic & Medicinal Chemistry, 16(22), 9817-9829 (2008)
A nickel (0) catalyzed cycloaddition of alkynes and isocyanates that affords pyrimidine-diones
Duong HA and Louie J
Tetrahedron, 62(32), 7552-7559 (2006)
Mario Lozanov et al.
Journal of the American Chemical Society, 124(10), 2106-2107 (2002-03-07)
A new two-step procedure for the synthesis of cyclohexenols has been developed. A nickel-catalyzed three-component addition of an enal, alkyne, and acetylenic tin affords substituted hept-4-en-6-ynals. The products of this first step then undergo a second nickel-catalyzed reaction with organozincs

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service