Skip to Content
Merck
All Photos(7)

Documents

275123

Sigma-Aldrich

Tributyl(1-ethoxyvinyl)tin

97%

Synonym(s):

(1-Ethoxyvinyl)tri-n-butylstannane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)3]3SnC(OC2H5)=CH2
CAS Number:
Molecular Weight:
361.15
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.476 (lit.)

bp

85-86 °C/0.1 mmHg (lit.)

density

1.069 g/mL at 25 °C (lit.)

SMILES string

CCCC[Sn](CCCC)(CCCC)C(=C)OCC

InChI

1S/C4H7O.3C4H9.Sn/c1-3-5-4-2;3*1-3-4-2;/h1,4H2,2H3;3*1,3-4H2,2H3;

InChI key

HGXJOXHYPGNVNK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This vinylstannane undergoes Stille coupling with a vinyl triflate, giving, after hydrolysis, an α,β−unsaturated ketone, thus acting as an acetyl anion equivalent.

Application

Electrophilic methyl ketone equivalent used in a recent synthesis of a 13-oxophorbine (chlorophyll) from the corresponding 13-bromochlorin.
Reagent employed in palladium-catalyzed coupling reactions with sulfoxides, α-haloethers, and chlorocyclobutenones. Also used to convert acid chlorides to α-oxygenated enones.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Journal of the American Chemical Society, 114, 1412-1412 (1992)
Tetrahedron Letters, 33, 4885-4885 (1992)
Tetrahedron Letters, 34, 2429-2429 (1993)
Younggi Choi et al.
The Journal of organic chemistry, 69(11), 3758-3764 (2004-05-22)
2-Azabicyclo[2.2.2]oct-5-enes bearing an endo alkenyl substituent were synthesized by Diels-Alder addition of methyl vinyl ketone to a 1,6-dihydropyridine derived from methyl nicotinate. Although 1,5-dienes with this skeleton were unreactive under thermal conditions, they were photochemically reactive. Irradiation of these dienes
Joydev K Laha et al.
The Journal of organic chemistry, 71(18), 7049-7052 (2006-08-26)
A new route to 13(1)-oxophorbines, the parent macrocycle of chlorophylls, begins with the synthesis of a 13-bromochlorin. Pd-mediated coupling of the latter with tributyl(1-ethoxyvinyl)tin and subsequent acidic hydrolysis afforded the 13-acetylchlorin (1). Treatment of 1 with NBS afforded the 15-bromo

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service