Skip to Content
Merck
All Photos(2)

Key Documents

496383

Sigma-Aldrich

1,10-Phenanthroline-5,6-dione

97%

Synonym(s):

Stahl phd oxidant, phd

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H6N2O2
CAS Number:
Molecular Weight:
210.19
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

mp

260 °C (dec.) (lit.)

functional group

ketone

SMILES string

O=C1C(=O)c2cccnc2-c3ncccc13

InChI

1S/C12H6N2O2/c15-11-7-3-1-5-13-9(7)10-8(12(11)16)4-2-6-14-10/h1-6H

InChI key

KCALAFIVPCAXJI-UHFFFAOYSA-N

General description

1,10-Phenanthroline-5,6-dione (phendio) forms Cu(II) and Ag(I) phendio complexes, which show potent anti-fungal and anti-cancer activity. The modification of glassy carbon (GC) electrodes with phendio complexes of transition metals leads to the catalytic oxidation of NADH at low overpotential.

Application

1,10-Phenanthroline-5,6-dione may be used in the preparation of homo- and heterometallic complexes with early transition metal ions.
A Bifunctional quinone oxidant which, when used in conjunction with Zn2+ catalysts, is used to affect the aerobic oxidation of secondary amines to a variety of value added motifs, including indoles.

Bioinspired Aerobic Oxidation of Secondary Amines and Nitrogen Heterocycles with a Bifunctional Quinone Catalyst

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xuyan Mao et al.
The Analyst, 136(2), 293-298 (2010-10-20)
To improve the electrocatalytic activities of carbon nanotubes (CNT) towards the oxidation of nicotinamide adenine dinucleotide (NADH), we derive them with a redox mediator, 1,10-phenanthroline-5,6-dione (PD), by the noncovalent functionalization method. The redox carbon nanotubes (PD/CNT/GC) show excellent electrocatalytic activities
Tetsuaki Fujihara et al.
Dalton transactions (Cambridge, England : 2003), (4)(4), 645-652 (2004-07-15)
Syntheses and pH dependent electrochemical properties of aqua-ruthenium(II) complexes, [Ru(trpy)(PDA-N,N')(OH2)](ClO4)2 ([1](ClO4)2) and [Ru(trpy)(PD-N,N')(OH2)](ClO4)2 ([2](ClO4)2) (trpy = 2,2':6',2''-terpyridine, PDA = 6-acetonyl-6-hydroxy-1,10-phenanthroline-5-one, PD = 1,10-phenanthroline-5,6-dione) are presented. Treatment of [Ru(trpy)(PD-N,N')Cl](PF6) with AgClO4 in a mixed solvent of acetone and H2O selectively produced
Aleksandra Pinczewska et al.
Journal of the American Chemical Society, 134(43), 18022-18033 (2012-10-11)
We report the combinatorial preparation and high-throughput screening of a library of modified electrodes designed to catalyze the oxidation of NADH. Sixty glassy carbon electrodes were covalently modified with ruthenium(II) or zinc(II) complexes bearing the redox active 1,10-phenanthroline-5,6-dione (phendione) ligand
Sudeshna Roy et al.
ChemMedChem, 3(9), 1427-1434 (2008-06-10)
Phenanthroline derivatives are of interest due to their potential activity against cancer, and viral, bacterial, and fungal infections. In a search for highly specific antitumor and antibacterial compounds, we report the activities of 1,10-phenanthroline-5,6-dione (phendione or L(1)), dipyrido[3,2-a:2',3'-c]phenazine (dppz or
E Lorenzo et al.
Biosensors & bioelectronics, 13(3-4), 319-332 (1998-06-27)
Various strategies based on the use of chemically modified electrodes for the development of amperometric biosensors are described. Particular emphasis is placed on materials capable of catalyzing the oxidation of NADH and coupling these with enzymatic activities for biosensor construction.

Related Content

he Stahl Lab focuses on the development of catalysts and catalytic reactions for selective oxidation of organic molecules, with particular emphasis on aerobic oxidation reactions.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service