Skip to Content
Merck
All Photos(2)

Key Documents

495336

Sigma-Aldrich

Poly(3-butylthiophene-2,5-diyl)

regioregular

Synonym(s):

P3BT

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23

mol wt

Mw 54,000 (typical)

color

black

solubility

chlorinated solvents: soluble (partially soluble in THF, diethylether)

fluorescence

λex 440 nm; λem 567 nm in chloroform

Mw/Mn

2.3 (typical)

SMILES string

[s]1c(c(cc1C)CCCC)C

InChI

1S/C10H16S/c1-4-5-6-10-7-8(2)11-9(10)3/h7H,4-6H2,1-3H3

InChI key

DUOSBQJOYVIVOR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Conducting polymer, 80-90% head-to-tail regiospecific conformation.
Poly(3-butylthiophene-2,5-diyl) (P3BT) is an alkylthiophene based conducting polymer that can be used as a donor molecule in the development of organic electronics. It is a π-conjugating polymer with a π-π stacking distance of 0.395 nm.

Application

P3BT can act as a hole transporting layer (HTL) which can potentially be used in the fabrication of organic field effect transistors (OFETs), chemical sensors, rechargeable batteries and polymeric solar cells (PSCs).
Rechargeable battery electrodes, electrochromic devices, chemical and optical sensors, light-emitting diodes, microelectrical amplifiers, field-effect transistors and non-linear optical materials.

Packaging

Packaged in glass bottles

Citation

For solid state properties see J. Am. Chem. Soc. .

Legal Information

Product of Rieke Metals, Inc.
Rieke is a registered trademark of Rieke Metals, Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

High molecular weights, polydispersities, and annealing temperatures in the optimization of bulk-heterojunction photovoltaic cells based on poly (3-hexylthiophene) or poly (3-butylthiophene)
Hiorns RC, et al.
Advances in Functional Materials, 16(17), 2263-2273 (2006)
Effect of alkyl side-chain length and solvent on the luminescent characteristics of poly (3-n-alkylthiophene)
Shin C and Lee H
Synthetic Metals, 140(2-3), 177-181 (2004)
All-organic thin-film transistors patterned by means of selective electropolymerization
Becker E, et al.
Applied Physics Letters, 83(19), 4044-4046 (2003)
Nanofiber preparation by whisker method using solvent-soluble conducting polymers
Samitsu S, et al.
Thin Solid Films, 516(9), 2478-2486 (2008)
Journal of the American Chemical Society, 117, 233-233 (1994)

Articles

The application of conducting polymers at the interface with biology is an exciting new trend in organic electronics research.

Intrinsically stretchable active layers for organic field-effect transistors (OFET) are discussed. Polymer structural modification & post-polymerization modifications are 2 methods to achieve this.

Novel Graphene‑Based Nanostructures Production, Functionalization, and Engineering

Novel Graphene‑Based Nanostructures Production, Functionalization, and Engineering

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service