Skip to Content
Merck
All Photos(1)

Key Documents

486728

Sigma-Aldrich

Hexane-1-13C

99 atom % 13C

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)413CH3
CAS Number:
Molecular Weight:
87.17
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

isotopic purity

99 atom % 13C

Assay

99% (CP)

refractive index

n20/D 1.375 (lit.)

bp

69 °C (lit.)

mp

−95 °C (lit.)

density

0.667 g/mL at 25 °C

mass shift

M+1

SMILES string

CCCCC[13CH3]

InChI

1S/C6H14/c1-3-5-6-4-2/h3-6H2,1-2H3/i1+1

InChI key

VLKZOEOYAKHREP-OUBTZVSYSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Central nervous system, Nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-14.8 °F - closed cup

Flash Point(C)

-26 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Abdul R Ahameethunisa et al.
Annals of clinical microbiology and antimicrobials, 11, 30-30 (2012-11-23)
Artemisia parviflora leaf extracts were evaluated for potential antimicrobial and antioxidant properties. Antimicrobial susceptibility assay was performed against ten standard reference bacterial strains. Antioxidant activity was analyzed using the ferric thiocyanate and 2, 2-Diphenyl-1-Picrylhydrazyl (DPPH) assays. Radical scavenging activity and
Takuya Miura et al.
Organic letters, 14(23), 6048-6051 (2012-11-21)
By applying a skeleton transformation reaction using dimethylsulfoxonium methylide, a novel reaction was identified by which 5,6,7,8-tetrahydrocoumarin with the electron-withdrawing group at C3 was led to the spirobicyclo[3.1.0]hexane-cyclohexane derivative. Moreover, by establishing the scope of this reaction, it was confirmed
Dong Min Jung et al.
Journal of food science, 77(12), C1249-C1255 (2012-10-31)
The chemical properties and oxidative stability of perilla oils obtained from roasted perilla seeds as affected by extraction methods (supercritical carbon dioxide [SC-CO(2)], mechanical press, and solvent extraction) were studied. The SC-CO(2) extraction at 420 bar and 50 °C and
Katharina Röttger et al.
The journal of physical chemistry. A, 117(12), 2469-2478 (2013-03-21)
Guanosine (G) derivatives in nonpolar aprotic solvents self-assemble to intricate hydrogen-bonded supramolecular architectures, including dimers, ribbons, and cyclic quartets. Considerable interest exists in the nature of the excited electronic states, their lifetimes and the radiationless deactivation mechanisms of the molecules
Monica Rosa Loizzo et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 53, 392-401 (2012-12-26)
The present study evaluates the influence of drying and cooking processes on the health properties of two bell Capsicum annuum L. cultivars Roggiano and Senise compared with fresh peppers. The content of phytochemicals decreased in the order fresh>dried>dried frying processes.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service