327077
Silver
flakes, 10 μm, ≥99.9% trace metals basis
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Linear Formula:
Ag
CAS Number:
Molecular Weight:
107.87
EC Number:
MDL number:
UNSPSC Code:
12141740
PubChem Substance ID:
NACRES:
NA.23
Recommended Products
Quality Level
Assay
≥99.9% trace metals basis
form
flakes
resistivity
1.59 μΩ-cm, 20°C
particle size
10 μm
bp
2212 °C (lit.)
mp
960 °C (lit.)
density
10.49 g/cm3 (lit.)
SMILES string
[Ag]
InChI
1S/Ag
InChI key
BQCADISMDOOEFD-UHFFFAOYSA-N
General description
Silver flakes (Ag) are micro-sized flakes with a high aspect ratio. They can be used as conductive fillers that result in high conductive devices at low volumetric concentrations. Silver flakes are cost-efficient fillers compared with conventional nanowires and nanoparticles.
Application
Ag flakes can be used in the synthesis of nanoparticles, which can be used for a variety of applications such as electronics, optoelectronics, and flexible electronics.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 1
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Printable elastic conductors by in situ formation of silver nanoparticles from silver flakes
Matsuhisa N, et al.
Nature Materials, 16(8), 834-834 (2017)
Enhancing the Performance of Stretchable Conductors for E-Textiles by Controlled Ink Permeation
Jin H, et al.
Advanced Materials, 29(21), 1605848-1605848 (2017)
Conductive and stretchable silver-polymer blend for electronic applications
Proceedings, 6(8), 812-816 (2016)
Printable elastic conductors with a high conductivity for electronic textile applications
Matsuhisa N, et al.
Nature Communications, 6(8), 7461-7461 (2015)
S S Sudha et al.
Indian journal of experimental biology, 51(5), 393-399 (2013-07-05)
Silver nanoparticles is known to have antimicrobial affects. Cyanobacteria isolates from muthupet mangrove includes Aphanothece sp, Oscillatoria sp, Microcoleus sp, Aphanocapsa sp, Phormidium sp, Lyngbya sp, Gleocapsa sp, Synechococcus sp, Spirulina sp with were set in compliance with their cellular
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service