Skip to Content
Merck
All Photos(2)

Key Documents

467006

Sigma-Aldrich

1-Ethynyl-2-fluorobenzene

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H4C≡CH
CAS Number:
Molecular Weight:
120.12
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.526 (lit.)

bp

150 °C (lit.)

density

1.06 g/mL at 25 °C (lit.)

functional group

fluoro

storage temp.

2-8°C

SMILES string

Fc1ccccc1C#C

InChI

1S/C8H5F/c1-2-7-5-3-4-6-8(7)9/h1,3-6H

InChI key

YFPQIXUNBPQKQR-UHFFFAOYSA-N

General description

1-Ethynyl-2-fluorobenzene, a fluorinated benzene derivative, is a terminal alkyne.

Application

1-Ethynyl-2-fluorobenzene has been employed in the cross-coupling of phenylacetylenes. It may be used in the preparation of 3-(2-deoxy-β-D-ribofuranosyl)-6-(2-fluorophenyl)-2,3-dihydrofuro-[2,3-d]pyrimidin-2-one and 4-(2-fluorophenylethynyl)-3-methyl-5-phenylisoxazole.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

98.6 °F - closed cup

Flash Point(C)

37 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and Biological Evaluation of Novel 1, 2, 3-Triazolonucleotides.
Glowacka IE, et al.
Arch. Pharm. (Weinheim), 346(4), 278-291 (2013)
Halophenyl furanopyrimidines as potent and selective anti-VSV agents.
McGuigan C, et al.
Antiviral Chem. Chemother., 14(3), 165-165 (2003)
Tetrahedron Letters, 35, 3689-3689 (1994)
Synthesis of Novel 1-Hydroxy-2-(1, 2, 3-triazol-1-yl) ethylphosphonates and 2-Hydroxy-3-(1, 2, 3-triazol-1-yl) propylphosphonates.
Glowacka IE, et al.
Phosphorus, Sulfur, and Silicon and the Related Elements, 186(3), 431-449 (2011)
Synthesis of 3, 4, 5-Trisubstituted Isoxazoles through Gold-Catalyzed Cascade Cyclization- Oxidative Alkynylation and Cyclization-Fluorination of 2-Alkynone O-Methyloximes.
Song D-H and Ryu J-S.
Bull. Korean Chem. Soc., 35(19), 2635-2644 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service