All Photos(1)
About This Item
Linear Formula:
HC≡CC6H4NO2
CAS Number:
Molecular Weight:
147.13
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
97%
mp
148-150 °C (lit.)
functional group
nitro
SMILES string
[O-][N+](=O)c1ccc(cc1)C#C
InChI
1S/C8H5NO2/c1-2-7-3-5-8(6-4-7)9(10)11/h1,3-6H
InChI key
GAZZTEJDUGESGQ-UHFFFAOYSA-N
Related Categories
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Novel cross-linkers for PDMS networks for controlled and well distributed grafting of functionalities by click chemistry.
Madsen FB, et al.
Polym. Chem., 4(5), 1700-1707 (2013)
Synthesis of telechelic vinyl/allyl functional siloxane copolymers with structural control.
Madsen FB, et al.
Polym. Chem., 5(24), 7054-7061 (2014)
Dipolar cross-linkers for PDMS networks with enhanced dielectric permittivity and low dielectric loss.
Madsen FB, et al.
Smart Materials and Structures, 22(10), 104002-104002 (2013)
Fabiola Zapata et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 21(27), 9797-9808 (2015-06-03)
Several bis(triazolium)-based receptors have been synthesized as chemosensors for anion recognition. The central naphthalene core features two aryltriazolium side-arms. NMR experiments revealed differences between the binding modes of the two triazolium rings: one triazolium ring acts as a hydrogen-bond donor
Mohammad Zafar et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 25(59), 13537-13546 (2019-07-25)
Building upon previous work, the chemistry of [(η6 -p-cymene)Ru{P(OMe)2 OR}Cl2 ], (R=H or Me) has been extended with [H2 B(mbz)2 ]- (mbz=2-mercaptobenzothiazolyl) using different Ru precursors and borate ligands. As a result, a series of 1,3-N,S-chelated ruthenium borate complexes, for
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service