Skip to Content
Merck
All Photos(1)

Key Documents

450782

Sigma-Aldrich

Chromium(II) chloride

anhydrous, powder, 99.99% trace metals basis

Synonym(s):

Chromium dichloride, Chromous chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CrCl2
CAS Number:
Molecular Weight:
122.90
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
anhydrous
form:
powder

grade

anhydrous

Assay

99.99% trace metals basis

form

powder

impurities

≤150.0 ppm Trace Metal Analysis

mp

824 °C (lit.)

density

2.9 g/mL at 25 °C (lit.)

SMILES string

Cl[Cr]Cl

InChI

1S/2ClH.Cr/h2*1H;/q;;+2/p-2

InChI key

XBWRJSSJWDOUSJ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Mediates the condensation of the aldehydes with trisubstituted chloroalkenes. Used in the synthesis of a highly active precatalyst for ethylene oligimerization.
The vapor-phase co-reductions with other metal halides by hydrogen results in finely divided intermetallics with applications as structural materials or compounds with useful thermoelectric, magnetic, and oxidation-resistance properties.
Used to make bis(indenyl)chromium dimer and related products.
Used with iodoform and tetraethyl ethylenediamine to convert terminal olefins into trans-iodocyclopropanes in excellent yields.

accessory

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Small, B.L., et al.
Macromolecules, 37, 4375-4375 (2004)
Journal of Organometallic Chemistry, 692, 520-520 (2007)
Falck, J.R., et al.
Tetrahedron Letters, 45, 3039-3039 (2004)
Heinemann, O. et al.
Organometallics, 15, 5462-5462 (1996)
Min J Kwon et al.
Life sciences, 87(13-14), 401-404 (2010-08-21)
it has been suggested that Chromium (Cr), one of the essential minerals, can be beneficial to type 2 diabetic patients because it lowers blood glucose levels by improving various steps in insulin action. A few studies reported that Cr might

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service