Skip to Content
Merck
All Photos(1)

Key Documents

449946

Sigma-Aldrich

Neodymium(III) chloride

anhydrous, powder, ≥99.99% trace metals basis

Synonym(s):

Neodymium chloride, Neodymium trichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NdCl3
CAS Number:
Molecular Weight:
250.60
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
anhydrous
form:
powder
Pricing and availability is not currently available.

grade

anhydrous

Assay

≥99.99% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: neodymium

impurities

≤100.0 ppm Trace Rare Earth Analysis

mp

784 °C (lit.)

density

4.134 g/mL at 25 °C (lit.)

SMILES string

Cl[Nd](Cl)Cl

InChI

1S/3ClH.Nd/h3*1H;/q;;;+3/p-3

InChI key

ATINCSYRHURBSP-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Application


  • Fluorine-free organic electrolytes for the stable electrodeposition of neodymium metal: This study investigates alternative electrolytes for neodymium metal deposition, aiming for more sustainable battery manufacturing processes (Geysens et al., 2023).

  • The use of artificial neural network (ANN) for modeling adsorption of sunset yellow onto neodymium modified ordered mesoporous carbon: Highlights the use of Neodymium(III) chloride in advanced adsorption processes for environmental applications, modeling the interactions using ANN (Ahmad et al., 2020).


Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dongyan Zhang et al.
Journal of inorganic biochemistry, 103(7), 935-939 (2009-06-06)
After treatment with DyCl(3) and NdCl(3), two strains of mutated bacilli showing increased alpha-amylase activity were isolated. The alpha-amylase genes were amplified, cloned, and sequenced. Sequencing revealed that there were either 11 or 14 bases altered in the two genes.
Yue-tao Yang et al.
Guang pu xue yu guang pu fen xi = Guang pu, 22(4), 566-568 (2003-08-27)
Photoacoustic(PA) phase-resolved method is used to separate the overlapping peaks of lanthanide complex for the first time. PA peaks of Nd3+ are covered completely by the PA absorption of tryptophan(Trp) for Nd(Trp)3Cl3.H2O within 300-400 nm. The patterns of PA phase
Yan Lu et al.
Carbohydrate research, 338(24), 2913-2919 (2003-12-12)
The single-crystal structure of neodymium chloride-ribopyranose pentahydrate, NdCl3.C5H10O5.5H2O was determined to have Mr=490.80, a=9.138(11), b=8.830(10), c=9.811(11) A, beta=94.087(18) degrees, V=789.7(16) A3, P2(1), Z=2, mu=0.71073 A and R=0.0198 for 2075 observed reflections. The ligand of the title complex was observed in
Antioxidant protection mechanism of chick bone marrow: effects of toxicant induced stress.
N Ghosh et al.
Indian journal of experimental biology, 25(4), 253-255 (1987-04-01)
N Ghosh et al.
Indian journal of experimental biology, 29(3), 226-229 (1991-03-01)
Acute single dose (ip) administration of two rare earth elements like lanthanum chloride (250 mg/kg body wt) and neodymium chloride (200 mg/kg body wt) to chicks have been found to reduce the activity of certain erythrocyte membrane bound enzymes, viz.

Articles

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service