Skip to Content
Merck
All Photos(2)

Key Documents

447420

Sigma-Aldrich

Orcinol

97%

Synonym(s):

5-Methylresorcinol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H3-1,3-(OH)2
CAS Number:
Molecular Weight:
124.14
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
solid
Assay:
97%

Quality Level

Assay

97%

form

solid

mp

106-112 °C (lit.)

SMILES string

Cc1cc(O)cc(O)c1

InChI

1S/C7H8O2/c1-5-2-6(8)4-7(9)3-5/h2-4,8-9H,1H3

InChI key

OIPPWFOQEKKFEE-UHFFFAOYSA-N

Application

Orcinol can be used to synthesize:
  • Orcinol-containing azacryptands for use in optical amplifiers and light-emitting devices.
  • Ternary co-crystal with 4,4′-bipyridine.
  • Low-density carbon aerogels in the presence of formaldehyde.
  • PEG-orcinol coumarins with potent tyrosinase inhibitory activity.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Preparation of carbon aerogels from 5-methylresorcinol-formaldehyde gels
Perez-Caballero F, et al.
Microporous and Mesoporous Materials : The Official Journal of the International Zeolite Association, 108(1-3), 230-236 (2008)
Nianxia Sun et al.
Plant foods for human nutrition (Dordrecht, Netherlands), 74(3), 376-382 (2019-06-22)
Water-extracted arabinoxylans (WEAXs) of different varieties and structures have important effects on wheat end products. However, the functional performances of WEAXs, particularly relating to prebiotic potential, are not yet clear. The present study compared the structural features, physicochemical properties, and
Dibenzofurans from the marine sponge-derived ascomycete Super1F1-09.
Rateb ME, et al.
Botanica Marina, 53(6), 499-506 (2010)
Yong-qi Tian et al.
Natural product research, 29(9), 820-826 (2014-12-30)
A new citromycetin analogue, ascomycotin A (1), together with eight known compounds, wortmannilactone E (2), orcinol (3), orsellinic acid (4), isosclerone (5), (3R,4S)-( - )-4-hydroxymellein (6), diorcinol (7), chaetocyclinone B (8) and 2,5-dimethoxy-3,6-di(p-methoxypheny1)-1,4-benzoquinone (9), was isolated from the fungal strain Ascomycota sp.
Goutam Ghosh et al.
Pharmacognosy research, 7(1), 110-113 (2015-01-20)
Clerodendrum viscosum is commonly found in India and Bangladesh. Previously, various parts of this plant were reported for treatment of different types of diseases and there was no report on GC-Ms analysis. To analyze and characterize the phytochemical compounds of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service